Skip to Content
Merck
All Photos(1)

Key Documents

8.01786

Sigma-Aldrich

Bromobenzene

for synthesis

Synonym(s):

Bromobenzene, Phenyl bromide

Sign Into View Organizational & Contract Pricing

Select a Size

5 ML
€25.00
250 ML
€40.40
1 L
€119.00
25 L
€2,070.00

€25.00


Estimated to ship on30 May 2025


Request a Bulk Order

Select a Size

Change View
5 ML
€25.00
250 ML
€40.40
1 L
€119.00
25 L
€2,070.00

About This Item

Empirical Formula (Hill Notation):
C6H5Br
CAS Number:
Molecular Weight:
157.01
MDL number:
UNSPSC Code:
12352101
EC Index Number:
203-623-8
NACRES:
NA.22

€25.00


Estimated to ship on30 May 2025


Request a Bulk Order

vapor pressure

4 hPa ( 20 °C)

Quality Level

Assay

≥99% (GC)

form

liquid

autoignition temp.

565 °C

potency

2383 mg/kg LD50, oral (Rat)

expl. lim.

0.5-2.5 % (v/v)

bp

156 °C/1013 hPa

mp

-31 °C

transition temp

flash point 51 °C

density

1.49 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

Brc1ccccc1

InChI

1S/C6H5Br/c7-6-4-2-1-3-5-6/h1-5H

InChI key

QARVLSVVCXYDNA-UHFFFAOYSA-N

Application

  • Synthesis of High-Quality Perovskite Nanocrystals: Utilizing bromobenzene for aliphatic nucleophilic substitution enabled the synthesis of high-quality cesium lead bromide perovskite nanocrystals, which are critical in optoelectronic applications (Li et al., 2019).
  • Noncatalytic Bromination of Benzene: This study demonstrated the necessary conditions for the noncatalytic bromination of benzene to produce bromobenzene, providing valuable information for industrial synthesis processes (Shernyukov et al., 2016).

Analysis Note

Assay (GC, area%): ≥ 99 %
Density (d 20 °C/ 4 °C): 1.494 - 1.496
Identity (IR): passes test

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Flam. Liq. 3 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

123.8 °F - closed cup

Flash Point(C)

51.0 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service