Skip to Content
Merck
All Photos(1)

Key Documents

8.00458

Sigma-Aldrich

p-Anisidine

for synthesis

Synonym(s):

p-Anisidine, 4-Methoxyaniline, 4-Aminoanisole, 4-Aminophenyl methyl ether

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
€44.20
500 G
€125.00

€44.20


Estimated to ship on27 May 2025


Request a Bulk Order

Select a Size

Change View
100 G
€44.20
500 G
€125.00

About This Item

Linear Formula:
4-(CH3O)C6H4NH2
CAS Number:
Molecular Weight:
123.15
MDL number:
UNSPSC Code:
12352116
EC Index Number:
203-254-2
NACRES:
NA.22

€44.20


Estimated to ship on27 May 2025


Request a Bulk Order

vapor pressure

0.02 hPa ( 20 °C)

Quality Level

Assay

≥98.0% (GC)

form

crystals

autoignition temp.

450 °C

pH

7.7 (20 °C, 1 g/L in H2O)

mp

55-58 °C

transition temp

flash point 122 °C

solubility

21 g/L

density

1.06 g/cm3 at 63 °C

bulk density

700 kg/m3

storage temp.

2-30°C

SMILES string

Nc1ccc(cc1)OC

InChI

1S/C7H9NO/c1-9-7-4-2-6(8)3-5-7/h2-5H,8H2,1H3

InChI key

BHAAPTBBJKJZER-UHFFFAOYSA-N

Application

  • Interfacing poly(p-anisidine) with photosystem I: This study details the integration of poly(p-anisidine) with photosystem I to develop photoactive composite films, highlighting a novel approach to enhance the functionality of biomaterials in energy conversion systems (Nabhan et al., 2024).
  • Quality assessment in food processing: Although focused on the drying processes of olive pomace, this research contributes to understanding quality management in food oils, which can be indirectly related to chemical studies like those involving p-Anisidine in quality control scenarios (Turan et al., 2023).
  • Electroactive material for solar cells: Research on N,N-Bis(9-methyl-3-carbazolyl)-4-anisidine demonstrates its potential as an electroactive material in perovskite solar cells, offering insights into the use of anisidine derivatives in advanced energy materials applications (Keruckas et al., 2023).

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Melting range (lower value): ≥ 55 °C
Melting range (upper value): ≤ 59 °C
Identity (IR): passes test

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Carc. 1B - STOT RE 2

Storage Class Code

6.1A - Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

251.6 °F - closed cup

Flash Point(C)

122 °C - closed cup


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

EU REACH Annex XVII (Restriction List)

CAS No.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service