Skip to Content
Merck
All Photos(1)

Key Documents

7610-OP

Millipore

Sodium Acetate

OmniPur® Grade, 99.0-101.0%, Trihydrate

Synonym(s):

Sodium acetate trihydrate, Acetic acid sodium salt trihydrate

Sign Into View Organizational & Contract Pricing

Select a Size

500 G
€43.30
5 KG
€275.00

€43.30


Estimated to ship on23 May 2025


Request a Bulk Order

Select a Size

Change View
500 G
€43.30
5 KG
€275.00

About This Item

Linear Formula:
CH3COONa · 3H2O
CAS Number:
Molecular Weight:
136.08
Beilstein:
3732037
MDL number:
UNSPSC Code:
12352302
NACRES:
NA.21

€43.30


Estimated to ship on23 May 2025


Request a Bulk Order

Quality Level

form

powder

autoignition temp.

1112 °F

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

shipped in

ambient

SMILES string

O.O.O.[Na+].CC([O-])=O

InChI

1S/C2H4O2.Na.3H2O/c1-2(3)4;;;;/h1H3,(H,3,4);;3*1H2/q;+1;;;/p-1

InChI key

AYRVGWHSXIMRAB-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description



Molecular biology grade.

Specifications:
Assay: 99.0-101.0%
Calcium, magnesium, and R2O3 precipitate: 0.01% max
Chloride (Cl): 0.001% max
DNase: None detected
Heavy metals (as Pb): 0.0005% max
Insoluble matter: 0.005% max
Iron (Fe): 0.0005% max
pH (5%, water): 7.5-9.2
Phosphate (PO4): 0.0005% max
Potassium (K): 0.005% max
Protease: None detected
RNase: None detected
Substances reducing permanganate: To pass test
Sulfate (SO4): 0.002% max





Intended for laboratory and manufacturing use only. Not for drug, food, or household use.

Other Notes

Enzyme substrate for acetate kinase (EC 2.7.2.1)

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
OMNIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service