Skip to Content
Merck
All Photos(2)

Key Documents

1.06267

Supelco

Sodium acetate trihydrate

for analysis EMSURE® ACS,ISO,Reag. Ph Eur

Synonym(s):

Acetic acid sodium salt trihydrate

Sign Into View Organizational & Contract Pricing

Select a Size

500 G
€42.10
1 KG
€64.80
5 KG
€227.00
12 KG
€746.00
50 KG
€2,110.00

€42.10


Estimated to ship on24 May 2025


Request a Bulk Order

Select a Size

Change View
500 G
€42.10
1 KG
€64.80
5 KG
€227.00
12 KG
€746.00
50 KG
€2,110.00

About This Item

Linear Formula:
CH3COONa · 3H2O
CAS Number:
Molecular Weight:
136.08
Beilstein:
3732037
MDL number:
UNSPSC Code:
12352107
EC Index Number:
204-823-8
NACRES:
NA.21

€42.10


Estimated to ship on24 May 2025


Request a Bulk Order

grade

ACS reagent

Quality Level

Agency

reag. ISO
reag. Ph. Eur.

product line

EMSURE®

Assay

99.0-101.0% (perchloric acid titration)
99.0-101.0% dry basis (perchloric acid titration)

form

solid

autoignition temp.

1112 °F
607 °C

potency

3530 mg/kg LD50, oral (Rat)
>10000 mg/kg LD50, skin (Rabbit)

impurities

≤0.005% Insoluble matter
≤0.005% Reducing substances

loss

39.0-40.5% loss on drying, 130 °C

pH

7.5-9.2 (20 °C, 50 g/L in H2O)

mp

58 °C

transition temp

flash point >250 °C (anhydrous substance)

solubility

613 g/L

density

1.42 g/cm3 at 20 °C

bulk density

900 kg/m3

anion traces

chloride (Cl-): ≤0.0005%
phosphate (PO43-): ≤0.0002%
sulfate (SO42-): ≤0.002%

cation traces

Al: ≤0.0005%
As: ≤0.0002%
Ca: ≤0.001%
Ca: ≤0.005%
Cu: ≤0.0003%
Fe: ≤0.0005%
K: ≤0.005%
Mg: ≤0.0005%
Mg: ≤0.005%
N: ≤0.001%
Pb: ≤0.0005%
heavy metals (as Pb): ≤0.0005%

UV absorption

λ: 254 nm Amax: ≤0.02
λ: 280 nm Amax: ≤0.01
λ: 350 nm Amax: ≤0.01

storage temp.

2-30°C

SMILES string

O.O.O.[Na+].CC([O-])=O

InChI

1S/C2H4O2.Na.3H2O/c1-2(3)4;;;;/h1H3,(H,3,4);;3*1H2/q;+1;;;/p-1

InChI key

AYRVGWHSXIMRAB-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application


  • Direct Affinity Ligand Immobilization onto Bare Iron Oxide Nanoparticles Enables Efficient Magnetic Separation of Antibodies: Highlights the role of sodium acetate trihydrate in bioengineering, particularly in the enhancement of magnetic nanoparticle performance for bioseparation applications (Zimmermann I, Berensmeier S, 2024).

  • Physicochemical and antioxidant properties of pectin fractions extracted from lemon (Citrus Eureka) peels: Details the use of sodium acetate trihydrate in the extraction and analysis of bioactive compounds, supporting its critical role in food science and nutraceutical applications (Su Z, Li G, 2024).

Other Notes

Enzyme substrate for acetate kinase (EC 2.7.2.1)

Linkage

Replaces: SX0255; SX0255-1; SX0255-3; SX0255-20

Analysis Note

Assay (perchloric acid titration): 99.0 - 101.0 %
Assay (perchloric acid titration, calculated on dried substance): 99.0 - 101.0 %
Identity: passes test
Appearance of solution: passes test
Insoluble matter: ≤ 0.005 %
pH-value (5 %; water): 7.5 - 9.0
Chloride (Cl): ≤ 0.0005 %
Phosphate (PO₄): ≤ 0.0002 %
Sulfate (SO₄): ≤ 0.002 %
Total nitrogen (N): ≤ 0.001 %
Heavy metals (as Pb): ≤ 0.0005 %
Al (Aluminium): ≤ 0.0005 %
As (Arsenic): ≤ 0.0002 %
Ca (Calcium): ≤ 0.001 %
Cu (Copper): ≤ 0.0003 %
Fe (Iron): ≤ 0.0005 %
K (Potassium): ≤ 0.005 %
Mg (Magnesium): ≤ 0.0005 %
Pb (Lead): ≤ 0.0005 %
Calcium and Magnesium (as Ca): ≤ 0.005 %
Substances reducing permanganate: passes test
Reducing substances: ≤ 0.005 %
Loss on drying (130 °C): 39.0 - 40.5 %
UV absorption (254 nm; 1 mol/l; 1 cm; water): ≤ 0.02
UV absorption (280 nm; 1 mol/l; 1 cm; water): ≤ 0.01
UV absorption (350 nm; 1 mol/l; 1 cm; water): ≤ 0.01
Corresponds to ACS,ISO,Reag. Ph Eur

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service