Skip to Content
Merck

71978

One Step RT-PCR Master Mix Kit

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.55
UNSPSC Code:
41106303
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment



Storage Class

10 - Combustible liquids

wgk

WGK 3



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library