Skip to Content
Merck
All Photos(1)

Key Documents

71712-M

Millipore

D-Tube96 Dialyzer, 6-8 kDa

Synonym(s):

D-Tube dialyzer

Sign Into View Organizational & Contract Pricing

Select a Size

96 EA
€566.00

€566.00


Estimated to ship on22 May 2025



Select a Size

Change View
96 EA
€566.00

About This Item

UNSPSC Code:
41104900
NACRES:
NB.24

€566.00


Estimated to ship on22 May 2025


manufacturer/tradename

Novagen®

Quality Level

storage condition

OK to freeze

storage temp.

10-30°C

Components

•: 1: D-Tube96 Dialyzer, 6-8 kDa


•: 96: D-Tube96 Dialyzer caps


•: 1: Aluminum Plate Sealer

Warning

Toxicity: Standard Handling (A)

Legal Information

NOVAGEN is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10-13 - German Storage Class 10 to 13


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service