Skip to Content
Merck
All Photos(1)

Key Documents

71509-M

Millipore

D-Tube Dialyzer Maxi, MWCO 6-8 kDa

Synonym(s):

D-Tube Maxi Dialyzer, MWCO 6-8 kDa Dialyzer, Maxi Dialyzer

Sign Into View Organizational & Contract Pricing

Select a Size

10 EA
€169.00

€169.00


Estimated to ship on30 May 2025



Select a Size

Change View
10 EA
€169.00

About This Item

UNSPSC Code:
41104900

€169.00


Estimated to ship on30 May 2025


manufacturer/tradename

Novagen®

Quality Level

storage condition

OK to freeze

storage temp.

10-30°C

Related Categories

General description


D-Tube Dialyzers Mini, Midi, Maxi, and Mega
The D-Tube Dialyzers be used for dialysis and electroelution of proteins, RNA, DNA, and oligonucleotides from polyacrylamide or agarose gels. The disposable, single-use tubes require no syringes, microcentrifuge, or laborious steps to manipulate small sample volumes. The sample is added and removed using a standard laboratory pipet. Available with molecular weight cutoffs (MWCO) from 3.5 to 14 kDa, the D-Tube Dialyzers are designed in four volume capacities: mini (10-250 µl), midi (50-800 micro;l), maxi (100-3000 µl), and mega (10ml, 15ml, and 20 ml). The membrane is ultra-clean, EDTA-treated regenerated cellulose, sulfur- and heavy metal-free.

The D-Tube Dialyzers Mini, Midi, and Maxi contain 10 D-Tube Dialyzers and one floating rack that can hold up to four D-Tube Dialyzers. The D-Tube Dialyzers Mega contain 10 or 50 tubes and one or 5 floating racks to hold one D-Tube Mega in the exchange buffer.

D-Tube Electroelution Accessory Kit
The D-Tube Electroelution Accessory Kit provides a support tray for the D-Tube Dialyzers (compatible with most commercially-available horizontal electrophoresis units) and optimized reagents for protein and nucleic acid precipitation following electroelution. The kit also contains an optimized buffer for the removal of SDS from extracted protein samples, making them compatible with MALDI-MS analysis.The combination of D-Tube Dialyzers and D-Tube Electroelution Accessory Kit provides a unique tool for extraction of any protein, protein-protein and protein-DNA complexes from non-denaturing and denaturing (SDS) polyacrylamide gels with 60% recovery yield in less than 2 hours. The procedure efficientlyrecovers proteins and simultaneously removes ampholytes from proteins run on 2D gels. Extracted proteins are compatible with most downstream applications such as MALDIMS, animal immunization for antibodyproduction, HPLC analysis, peptide mapping, and functional assays. In addition, D-Tube Dialyzers can be used for oligonucleotides, RNA and DNA extraction from both polyacrylamide and agarose gels. Efficientextraction (>90%) is achieved for 15 nt oligos and for up to 80 kbp DNA fragments.

Features and Benefits

  • Easy-to-handle dialyzers for buffer exchange and removal of urea, detergents
  • Ideal for electroelution of proteins, protein-DNA complexes, oligonucleotides, DNA, and RNA from polyacrylamide and agarose gels
  • One-step procedure that does not require syringes or any special equipment
  • Sample volume recovery >97%
  • Protease, RNAse, DNAse, and PCR product free

Warning

Toxicity: Standard Handling (A)

Legal Information

NOVAGEN is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service