Skip to Content
Merck
All Photos(1)

Key Documents

71194-M

Millipore

0.5 M THP Solution

Reducing agent compatible with His•Bind chromatography

Synonym(s):

Tris(3-hydroxypropyl)phosphine

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
€62.00
5 X 1 ML
€226.00

€62.00


Estimated to ship on13 May 2025



Select a Size

Change View
1 ML
€62.00
5 X 1 ML
€226.00

About This Item

UNSPSC Code:
41106511
NACRES:
NA.77

€62.00


Estimated to ship on13 May 2025


Quality Level

form

solution

manufacturer/tradename

Novagen®

storage condition

OK to freeze

technique(s)

protein purification: suitable

application(s)

sample preparation

storage temp.

−20°C

General description

Reducing agent compatible with HIS-BIND® chromatography
THP [Tris(hydroxypropyl)phosphine; C9H21O3P] is a water-soluble, odorless, sulfhydryl reducing agent that is more stable and effective than dithiothreitol (DTT). Against oxidized glutathione, the reducing capacity of THP (0.232 meq/g) is greater than that of TCEP (0.182 meq/g). THP is also more resistant to air oxidation than DTT and is compatible with immobilized metal affinity chromatography with HIS-BIND® and Ni-NTA His•Bind resins at concentrations up to 1.0 mM. The common working concentration of THP for many applications is 15 mM. THP is provided as a ready-to-use 0.5 M solution in water.

Purity: >80% by NMR. CAS: 4706-17-6. MW: 208.2.

Features and Benefits

Compatability with common methods for purifying His-tagged proteins - immobilized metal affinity chromatography with HIS-BIND® and Ni-NTA His•Bind resins at concentrations up to 1.0 mM

Stability: More stable and effective than traditional reducing agents like dithiothreitol (DTT)

Reducing Capacity: greater reducing capacity than TCEP against oxidized glutathione

Ready-to-use: THP is provided as a ready-to-use 0.5 M solution in water, making it easy to use in many applications with flexible usage

Warning

Toxicity: Corrosive (L)

Other Notes

Due to the nature of the Hazardous Materials in this shipment, additional shipping charges may be applied to your order. Certain sizes may be exempt from the additional hazardous materials shipping charges. Please contact your local sales office for more information regarding these charges.

Legal Information

HIS-BIND is a registered trademark of Merck KGaA, Darmstadt, Germany
NOVAGEN is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service