Skip to Content
Merck
All Photos(1)

Documents

71071

Sigma-Aldrich

pET-41 Ek/LIC Vector Kit - Novagen

Novagen′s pET-41 series is designed for cloning and high-level expression of peptide sequences fused with the 220 aa GST-Tag protein.

Synonym(s):

Ek/LIC Cloning Kits, pET-41 Ek/LIC Vector, pET-41 series

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106610
NACRES:
NA.54

Quality Level

manufacturer/tradename

Novagen®

storage condition

OK to freeze

shipped in

dry ice

storage temp.

−70°C

General description

Directional PCR cloning into the most powerful E. coli expression vectors

The pET-41 series is designed for cloning and high-level expression of peptide sequences fused with the 220 aa GST•Tag protein. Vector encoded sequence can be completely removed by cleaving the GST fusion protein with enterokinase. Note that the sequence is numbered by the pBR322 convention, so the T7 expression region is reversed on the vector map (TB317). The f1 origin is oriented so that infection with helper phage will produce virions containing single-stranded DNA that corresponds to the coding strand.

Warning

Toxicity: Multiple Toxicity Values, refer to MSDS

Legal Information

NOVAGEN is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Commercial use of this Product requires a commercial license from EMD Millipore Corporation. Commercial use shall include but not be limited to (1) use of the Product or its components in manufacturing; (2) use of the Product or its components to provide a service, information, or data to others in exchange for consideration; (3) use of the Product or its components (or any derivatives thereof) for therapeutic, diagnostic or prophylactic purposes (including as part of a device, chip, assay or other product); or (4) resale of the Product or its components, whether or not such Product or its components are resold for use in research. Nothing contained herein shall be deemed to represent or warrant that additional third party rights are not required for use of this Product.

This product contains genetically modified organisms (GMO). Within the EU GMOs are regulated by Directives 2001/18/EC and 2009/41/EC of the European Parliament and of the Council and their national implementation in the member States respectively. This legislation obliges MilliporeSigma to request certain information about you and the establishment where the GMOs are being handled. Clickhere for Enduser Declaration (EUD) Form.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service