Skip to Content
Merck
All Photos(1)

Key Documents

70911

Sigma-Aldrich

Origami B Competent Cell Set - Novagen

Escherichia coli, rod shaped

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106202
NACRES:
NA.81
Pricing and availability is not currently available.

Product Name

Origami B Competent Cell Set - Novagen,

biological source

Escherichia coli

Quality Level

manufacturer/tradename

Novagen®

storage condition

OK to freeze

growth mode

adherent or suspension

morphology

rod shaped

technique(s)

microbiological culture: suitable

shipped in

dry ice

storage temp.

−70°C

General description

Origami B host strains carry the same trxB/gor mutations as the original Origami strains, except that they are derived from a lacZY mutant of BL21. Thus the Origami B strains combine the desirable characteristics of BL21, Tuner and Origami hosts in one strain background. The trxB and gor mutations are selectable on kanamycin and tetracycline, respectively; therefore, this strain is recommended for use with pET plasmids carrying the ampicillin resistance marker bla.

DE3 indicates that the host is a lysogen of λDE3, and therefore carries a chromosomal copy of the T7 RNA polymerase gene under control of the lacUV5 promoter. Such strains are suitable for production of protein from target genes cloned in pET vectors by induction with IPTG.

pLysS strains express T7 lysozyme, which further suppresses basal expression of T7 RNA polymerase prior to induction, thus stabilizing pET recombinants encoding target proteins that affect cell growth and viability.
Genotypes:

Origami B

F-ompT hsdSB(rB- mB-) gal dcm lacY1 ahpC gor522::Tn10 trxB (KanR, TetR)

Origami B(DE3)

F-ompT hsdSB(rB- mB-) gal dcm lacY1 ahpC (DE3) gor522::Tn10 trxB (KanR, TetR)

Origami B(DE3)pLysS

F-ompT hsdSB(rB- mB-) gal dcm lacY1 ahpC (DE3) gor522::Tn10 trxB pLysS (CamR, KanR, TetR)

This product contains genetically modified organisms (GMO). Within the EU GMOs are regulated by Directives 2001/18/EC and 2009/41/EC of the European Parliament and of the Council and their national implementation in the member States respectively. This legislation obliges us to request certain information about you and the establishment where the GMOs are being handled. Click here for Enduser Declaration (EUD) Form.

Components

•2 x 0.2 mlOrigami B Competent Cells

•2 x 0.2 mlOrigami B(DE3) Competent Cells

•2 x 0.2 mlOrigami B(DE3)pLysS Competent Cells

•4 x 2 mlSOC Medium

•2 ngTest Plasmid

Warning

Toxicity: Multiple Toxicity Values, refer to MSDS (O)

Legal Information

NOVAGEN is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service