Skip to Content
Merck
All Photos(1)

Documents

70836-M

Sigma-Aldrich

Origami B Competent Cells - Novagen

Escherichia coli, rod shaped

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106202
NACRES:
NA.81

product name

Origami B Competent Cells - Novagen, Origami B host strains carry the same mutations as the original Origami strain, except that they are derived from a lacZY mutant of BL21 to enable precise control of expression levels using IPTG.

biological source

Escherichia coli

Quality Level

manufacturer/tradename

Novagen®

storage condition

OK to freeze

growth mode

adherent or suspension

morphology

rod shaped

technique(s)

microbiological culture: suitable

cell transformation

transformation efficiency: >2×106 cfu/μg

shipped in

dry ice

storage temp.

−70°C

General description

Genotype: F-ompT hsdSB(rB- mB-) gal dcm lacY1 ahpC gor522::Tn10 trxB (KanR, TetR)





This product contains genetically modified organisms (GMO). Within the EU GMOs are regulated by Directives 2001/18/EC and 2009/41/EC of the European Parliament and of the Council and their national implementation in the member States respectively. This legislation obliges us to request certain information about you and the establishment where the GMOs are being handled. Click here for Enduser Declaration (EUD) Form.
Origami B host strains carry the same trxB/gor mutations as the original Origami strains, except that they are derived from a lacZY mutant of BL21. Thus the Origami B strains combine the desirable characteristics of BL21, Tuner and Origami hosts in one strain background. The trxB and gor mutations are selectable on kanamycin and tetracycline, respectively; therefore, these strains are recommended for use only with pET plasmids carrying the ampicillin resistance marker bla.
Origami B host strains carry the same mutations as the original Origami strain, except that they are derived from a lacZY mutant of BL21 to enable precise control of expression levels using IPTG.

Components

0.4 ml1 mlComponent

•2 × 0.2 ml5 × 0.2 mlOrigami B Competent Cells

•2 × 2 ml4 × 2 mlSOC Medium

•10 µl10 µlTest Plasmid

Warning

Toxicity: Multiple Toxicity Values, refer to MSDS (O)

Legal Information

NOVAGEN is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service