70765-M
pET Expression System 22b - Novagen
pET vectors plus host strains with induction control
Synonym(s):
pET-22b(+) Vector
About This Item
Recommended Products
Quality Level
manufacturer/tradename
Novagen®
storage condition
OK to freeze
avoid repeated freeze/thaw cycles
shipped in
dry ice
wet ice
storage temp.
−70°C
General description
pET Expression Systems and pET Expression Systems plus Competent Cells provide core reagents needed for target gene cloning and expression
Components
Components for pET Expression Systems are similar for all systems unless otherwise stated with the specific pET Expression System description. pET Expression Systems include:
- 10 µg pET vector DNA (for each indicated plasmid)
- 0.2 ml BL21 Glycerol Stock
- 0.2 ml BL21(DE3) Glycerol Stock
- 0.2 ml BL21(DE3)pLysS Glycerol Stock
- 0.2 ml Induction Control Glycerol Stock
pET Expression Systems plus Competent Cellscontain all of the components of the specific pET Expression System, as well asthe following additional components, unless otherwise stated with the specificpET Expression System description:
- 0.2 ml NovaBlue Competent Cells
- 0.2 ml BL21(DE3) Competent Cells
- 0.2 ml BL21(DE3)pLysS Competent Cells
- 2 × 0.2 ml SOC Medium
- 10 µl Test Plasmid
Warning
Other Notes
This product contains genetically modified organisms (GMO). Within the EU GMOs are regulated by Directives 2001/18/EC and 2009/41/EC of the European Parliament and of the Council and their national implementation in the member States respectively. This legislation obliges MilliporeSigma to request certain information about you and the establishment where the GMOs are being handled. Click here for Enduser Declaration (EUD) Form.
Legal Information
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service