70527
100 mM IPTG Solution - Novagen
Convenient, ready-to-use solution
Synonym(s):
100 mM IPTG Solution - Novagen, IPTG Solution, 100 mM, Isopropyl -β-D-1-thiogalactopyranoside, Isopropylthio -β-D-galactoside
Select a Size
€138.00
Estimated to ship on25 May 2025
Select a Size
About This Item
€138.00
Estimated to ship on25 May 2025
Recommended Products
Quality Level
form
solution
mol wt
238.30 g/mol
manufacturer/tradename
Novagen®
storage condition
OK to freeze
avoid repeated freeze/thaw cycles
shipped in
wet ice
storage temp.
−20°C
SMILES string
S([C@@H]1O[C@@H]([C@@H]([C@@H]([C@H]1O)O)O)CO)C(C)C
InChI
1S/C9H18O5S/c1-4(2)15-9-8(13)7(12)6(11)5(3-10)14-9/h4-13H,3H2,1-2H3/t5-,6+,7+,8-,9+/m1/s1
InChI key
BPHPUYQFMNQIOC-NXRLNHOXSA-N
Application
Biochem/physiol Actions
Warning
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service