Skip to Content
Merck
All Photos(1)

Key Documents

69069

Millipore

Cleavage Control Protein

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
€110.00

€110.00


Estimated to ship on21 May 2025



Select a Size

Change View
10 μG
€110.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.54

€110.00


Estimated to ship on21 May 2025


form

liquid

Quality Level

manufacturer/tradename

Novagen®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

shipped in

wet ice

storage temp.

−20°C

General description

The Cleavage Control Protein is used to monitor performance of either thrombin or enterokinase cleavage conditions. The 48 kDa protein is cleaved into 2 fragments or cleaved by , which are easily visualized by SDS-PAGE. Thrombin cleavage results in fragments of 35 kDa and 13 kDa, and Enterokinase cleavage results in fragments of 32 kDa and 16 kDA.

Warning

Toxicity: Irritant (B)

Legal Information

NOVAGEN is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service