Skip to Content
Merck
All Photos(2)

Key Documents

69067-M

Millipore

Enterokinase Cleavage Capture Kit

Sign Into View Organizational & Contract Pricing

Select a Size

1 KIT
€448.00

€448.00


Estimated to ship on19 May 2025


Request a Bulk Order

Select a Size

Change View
1 KIT
€448.00

About This Item

UNSPSC Code:
41116133
NACRES:
NA.54

€448.00


Estimated to ship on19 May 2025


Request a Bulk Order

Packaging

Plastic ampoule

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Find highly-specific cleavage enzymes for isolating proteins during fusion protein purification including enterokinase, factor Xa, HRV 3C protease, and thrombin.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service