Skip to Content
Merck
All Photos(1)

Key Documents

676950-M

Sigma-Aldrich

Veratridine

≥98% (TLC), solid, Na⁺ channel opener, Calbiochem

Synonym(s):

Veratridine

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€114.00

€114.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€114.00

About This Item

Empirical Formula (Hill Notation):
C36H51NO11
CAS Number:
Molecular Weight:
673.79
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€114.00


Check Cart for Availability

Request a Bulk Order

Product Name

Veratridine, A Na+ channel opener.

Quality Level

description

Merck USA index - 14, 9954

Assay

≥98% (TLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white to yellow

solubility

DMSO: 25 mg/mL
ethanol: 25 mg/mL

storage temp.

2-8°C

SMILES string

N21[C@H]([C@]([C@@]3([C@H]([C@]4([C@]([C@H]5[C@]6(O[C@]7([C@H]([C@@]6(CC[C@@H]7OC(=O)c8cc(c(cc8)OC)OC)C)CC5)O)C4)(C[C@@H]3O)O)O)C2)O)(O)C)CC[C@@H](C1)C

InChI

1S/C36H51NO11/c1-19-6-11-26-31(3,40)35(43)25(17-37(26)16-19)33(42)18-34-24(32(33,41)15-27(35)38)10-9-23-30(34,2)13-12-28(36(23,44)48-34)47-29(39)20-7-8-21(45-4)22(14-20)46-5/h7-8,14,19,23-28,38,40-44H,6,9-13,15-18H2,1-5H3/t19-,23-,24-,25-,26-,27-,28-,30-,31+,32+,33+,34+,35-,36-/m0/s1

InChI key

FVECELJHCSPHKY-YFUMOZOISA-N

General description

A Na+ channel opener. Increases intracellular Ca2+ levels without affecting Na+ - Ca2+ exchange. Acts at neurotoxin receptor site 2 and preferentially binds to activated Na+ channels causing persistent activation. Induces cell death in bovine chromaffin cells.

Biochem/physiol Actions

Primary Target
Na+ channel

Warning

Toxicity: Highly Toxic (H)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Maroto, R., et al. 1994. Eur. J. Pharmacol. 270, 331.
Plattner, H., et al. 1994. J. Membr. Biol. 142, 229.
Rando, T.A., et al. 1989. J. Gen. Physiol.93, 43.
Rando, T.A., et al. 1986. Mol. Pharmacol.29, 467.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 1 Inhalation - Acute Tox. 1 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service