Skip to Content
Merck
All Photos(1)

Key Documents

6500-OP

Millipore

PBS Tablets

tablet (concave), OmniPur® Grade

Synonym(s):

PBS, Phosphate Buffered Saline, Phosphate Buffered Saline Tablets

Sign Into View Organizational & Contract Pricing

Select a Size

200 TABS
Inquire


Estimated to ship on23 May 2025


Request a Bulk Order

Select a Size

Change View
200 TABS
Inquire

About This Item

UNSPSC Code:
12161700
NACRES:
NA.25


Estimated to ship on23 May 2025


Request a Bulk Order

Product Name

PBS Tablets, OmniPur® Grade, Ideal for RNA electrophoresis in agarose

Quality Level

form

tablet (concave)

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

shipped in

ambient

General description

Each tablet makes a 1X solution when dissolved in 100 ml distilled or deionized water. If higher concentrations are desirable dissolve 2, 3 or 4 tablets per 100 m water. Molecular Biology Grade.

DNAse: none detected
pH (1 tablet/100 ml): 7.3-7.5
Protease: none detected
RNAse: none detected

Pack sizes:
6500: 1 PACK = 100 tablets
6501: 1 PACK = 200 tablets





Intended for laboratory and manufacturing use only. Not for drug, food, or household use.

Application


  • Discovering Hair Biomarkers of Alzheimer′s Disease Using High Resolution Mass Spectrometry-Based Untargeted Metabolomics.: The study uses PBS tablets in the sample preparation process for mass spectrometry, ensuring the accuracy and reliability of metabolomic profiling to identify biomarkers for Alzheimer′s disease (Su et al., 2023).

  • A systematic evaluation of quenching and extraction procedures for quantitative metabolome profiling of HeLa carcinoma cell under 2D and 3D cell culture conditions.: This article highlights the use of PBS tablets for cell washing and metabolite extraction, demonstrating their essential role in consistent and reproducible metabolomic analyses under varying cell culture conditions (Wang et al., 2023).

  • In situ digestion of alcohol-fixed cells for quantitative proteomics.: PBS tablets were employed in this study for washing and maintaining cells during in situ digestion, critical for accurate quantitative proteomics analysis and data integrity (Hatano et al., 2023).

Warning

Toxicity: Irritant (B)

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
OMNIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. Is this product sterile/usable in the context of microbiological analysis ?

    1 answer
    1. Product 6500-OP is not sterile and has not been tested for suitability for microbiological use.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service