Skip to Content
Merck
All Photos(1)

Key Documents

6506-OP

Millipore

Phosphate Buffered Saline

sterile, 10X, liquid, OmniPur® Grade

Synonym(s):

PBS, Phosphate Buffered Saline, 10X, Sterile

Sign Into View Organizational & Contract Pricing

Select a Size

4 L
Inquire


Estimated to ship on23 May 2025


Request a Bulk Order

Select a Size

Change View
4 L
Inquire

About This Item

UNSPSC Code:
12161700
NACRES:
NA.25


Estimated to ship on23 May 2025


Request a Bulk Order

Product Name

Phosphate Buffered Saline, OmniPur® Grade, 10X Liquid Concentrate, Sterile

Quality Level

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

clear colorless

shipped in

ambient

General description

Each liter makes 10 L of 1X PBS Buffer. Molecular Biology Grade.

DNAse: none detected
pH (10%, water) at 25°C: 7.3-7.5
RNAse: none detected
Sterility: Passes test





Intended for laboratory and manufacturing use only. Not for drug, food, or household use.

Application


  • Polyvinylalcohol Composite Filled with Carbon Dots Produced by Laser Ablation in Liquids.: This study explores the synthesis of polyvinyl alcohol (PVA) composites filled with carbon dots (CDs) using laser ablation in liquids (LAL). The composites were characterized and tested for potential biomedical applications, demonstrating enhanced mechanical properties and biocompatibility when evaluated in phosphate-buffered saline (PBS) solution (Cutroneo et al., 2024).

  • Exploring the Interplay between Tribocorrosion and Surface Chemistry of the ASTM F139 Surgical Stainless Steel in Phosphate-Buffered Saline Solution.: This research investigates the tribocorrosion behavior and surface chemistry of ASTM F139 surgical stainless steel in PBS. The study provides insights into the material′s performance and degradation mechanisms under physiological conditions, which is critical for biomedical implant applications (Macedo et al., 2024).

  • Biological Evaluation of Oral Care Products Using 3D Tissue-Engineered In Vitro Models of Plaque-Induced Gingivitis.: This study evaluates the efficacy of various oral care products using 3D tissue-engineered models of plaque-induced gingivitis. PBS was used as a control to simulate physiological conditions during the assessment of the anti-inflammatory and antimicrobial properties of the products (Barker et al., 2024).

  • UCST-Type Thermoresponsive Sol-Gel Transition Triblock Copolymer Containing Zwitterionic Polymer Blocks.: The research focuses on the development of a novel upper critical solution temperature (UCST) thermoresponsive sol-gel transition triblock copolymer. The material′s properties were extensively studied in PBS to evaluate its potential for drug delivery and other biomedical applications (Kawamura et al., 2024).

  • The Tardigrade-Derived Mitochondrial Abundant Heat Soluble Protein Improves Adipose-Derived Stem Cell Survival against Representative Stressors.: This study examines the protective effects of a tardigrade-derived protein on adipose-derived stem cells under stress conditions. The cells′ viability and function were assessed in PBS, providing valuable insights into the protein′s potential for enhancing stem cell therapies (Rolsma et al., 2024).

Warning

Toxicity: Irritant (B)

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
OMNIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service