Skip to Content
Merck
All Photos(1)

Key Documents

648492

Sigma-Aldrich

TRPM4 inhibitor, 9-Phenanthrol

The TRPM4 inhibitor, 9-Phenanthrol, also referenced under CAS 484-17-3, controls the biological activity of TRPM4. This small molecule/inhibitor is primarily used for Biochemicals applications.

Synonym(s):

TRPM4 inhibitor, 9-Phenanthrol, Phenanthren-9-ol, 9-Hydroxyphenanthrene

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
€96.90

€96.90


Estimated to ship on17 May 2025


Request a Bulk Order

Select a Size

Change View
50 MG
€96.90

About This Item

Empirical Formula (Hill Notation):
C14H10O
CAS Number:
Molecular Weight:
194.23
MDL number:
UNSPSC Code:
51111800
NACRES:
NA.77

€96.90


Estimated to ship on17 May 2025


Request a Bulk Order

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

brown

solubility

DMSO: 50 mg/mL

shipped in

wet ice

storage temp.

2-8°C

SMILES string

Oc1c2c(c3c(c1)cccc3)cccc2

InChI

1S/C14H10O/c15-14-9-10-5-1-2-6-11(10)12-7-3-4-8-13(12)14/h1-9,15H

InChI key

DZKIUEHLEXLYKM-UHFFFAOYSA-N

General description

A cell permeable hydroxytricyclic derivative that acts as a specific and reversible inhibitor of transient receptor potential melastatin 4 (TRPM4) (IC50 = 20 µM in HEK293 cells). Does not affect TRPM5 and cystic fibrosis transmembrane conductance regulators (CFTR) channels. Decreases the occurrence of early after depolarizations (EAD) and exhibits an anti-arrhythmic effect that is independent of the action of protein kinase A. Shown to abolish NMDA-induced burst firing in dopaminergic neurons. Does not affect K+ currents at ~10 µM, but at higher concentration (~100 µM) it reversibly reduces K+ currents.

Packaging

Packaged under inert gas

Warning

Toxicity: Irritant (B)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Grand, T., et al. 2008. Brit. Journ. Pharm.153, 1697.
Simard, C., 2012. Brit. Journ. Pharm.165, 2354.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service