Skip to Content
Merck
All Photos(2)

Key Documents

389412

Sigma-Aldrich

N-Methylmaleimide

97%

Synonym(s):

1-Methyl-1H-pyrrole-2,5-dione, 1-Methyl-2,5-dihydro-1H-pyrrole-2,5-dione, 1-Methylpyrrole-2,5-dione, N-Methyl maleic imide, N-Methylpyrrole-2,5-dione

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€82.40
25 G
€151.00

€82.40


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€82.40
25 G
€151.00

About This Item

Empirical Formula (Hill Notation):
C5H5NO2
CAS Number:
Molecular Weight:
111.10
Beilstein:
108550
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€82.40


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

form

powder

mp

94-96 °C (lit.)

functional group

imide
maleimide

SMILES string

CN1C(=O)C=CC1=O

InChI

1S/C5H5NO2/c1-6-4(7)2-3-5(6)8/h2-3H,1H3

InChI key

SEEYREPSKCQBBF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

N-Methylmaleimide can be used:
  • In the synthesis of organic structure directing agents for designing silicogermanate zeolites.[1]
  • As a dienophile in Diels-Alder reaction.[2]
  • For the synthesis of biologically active pyrrolo[2,1-a]isoquinolines.[3]
  • As a dipolarophile in [3 + 2] dipolar addition reactions.[4]
  • As a co-monomer in atom transfer radical polymerization of styrene to obtain styrene copolymers with programmed microstructure.[5]

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The ITQ-37 mesoporous chiral zeolite.
Sun J, et al.
Nature, 458(7242), 1154-1154 (2009)
Visible-light-induced oxidation/[3+ 2] cycloaddition/oxidative aromatization sequence: a photocatalytic strategy to construct pyrrolo [2, 1-a] isoquinolines.
Zou YQ, et al.
Angewandte Chemie (International Edition in English), 50(31), 7171-7175 (2011)
Asymmetric organocatalytic three-component 1, 3-dipolar cycloaddition: Control of stereochemistry via a chiral br?nsted acid activated dipole.
Chen XH, et al.
Journal of the American Chemical Society, 130(17), 5652-5653 (2008)
A facile procedure for controlling monomer sequence distribution in radical chain polymerizations.
Pfeifer S and Lutz JF
Journal of the American Chemical Society, 129(31), 9542-9543 (2007)
Hui Zhou et al.
The Analyst, 143(10), 2390-2396 (2018-04-27)
Glutathione (GSH) exhibits many cellular functions in human pathologies. A sensitive and simple method capable of assaying GSH would be useful to understand the mechanism of GSH-related diseases. In this study, a new colorimetric and fluorescent off-on probe, 3-oxo-3H-phenoxazin-7-ylthiophene-2-carboxylate, is

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service