Skip to Content
Merck
All Photos(1)

Key Documents

6.17650

Sigma-Aldrich

Water

HPLC grade

Synonym(s):

Water

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2O
CAS Number:
Molecular Weight:
18.02
Beilstein:
2050024
MDL number:
UNSPSC Code:
12191602
grade:
HPLC grade
technique(s):
HPLC: suitable
application(s):
general analytical
bp:
100 °C (lit.)

grade

HPLC grade

Quality Level

form

liquid

technique(s)

HPLC: suitable

refractive index

n20/D 1.34 (lit.)

pH

6.0-8.0 (25 °C, (77 °F))

bp

100 °C (lit.)

mp

0 °C (lit.)
0.0  °C (32.0 °F)

solubility

water: miscible

density

1.000 g/mL at 3.98 °C (lit.)

application(s)

general analytical

SMILES string

O

InChI

1S/H2O/h1H2

InChI key

XLYOFNOQVPJJNP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

HPLC-grade water is primarily used as a mobile phase component in the high-performance liquid chromatography (HPLC) method used for the separation of polar compounds.

Application

Water can be used as a mobile phase component:
  • For the analysis of water-soluble vitamins.
  • In preparing the mobile phase for the HPLC-ELSD analysis.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

No data available

Flash Point(C)

No data available


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Evaluation of various HILIC materials for the fast separation of polar compounds
Chauve B, et al.
Journal of Separation Science, 33(6-7), 752-764 (2010)
Application of parallel gradient high performance liquid chromatography with ultra-violet, evaporative light scattering and electrospray mass spectrometric detection for the quantitative quality control of the compound file to support pharmaceutical discovery
Squibb AW, et al.
Journal of Chromatography A, 1189(1-2), 101-108 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service