Skip to Content
Merck
All Photos(1)

Key Documents

616455

Sigma-Aldrich

TGF-β1, Human, Recombinant, CHO Cell Line

Synonym(s):

rhTGF-β1, Transforming Growth Factor-β1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202

Assay

≥97% (SDS-PAGE)

Quality Level

form

lyophilized

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

impurities

≤1 EU/μg Endotoxin (EU/μg TGF-β1)

shipped in

ambient

storage temp.

−20°C

General description

Recombinant, human transforming growth factor-β1 expressed in CHO cells. Promotes apoptosis in resting human B lymphocytes, glioma cells, and trigeminal neurinomal cells. Induces the accumulation of extracellular matrix in various diseases.

Biochem/physiol Actions

ED₅₀ = 0.04-0.1 ng/ml

Warning

Toxicity: Standard Handling (A)

Physical form

Lyophilized from a sterile filtered solution in 50 µg BSA/µg TGF-β1, 35% acetonitrile, 0.1% TFA.

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Avoid freeze/thaw cycles of solutions. Stock solutions are stable for up to 3 months at -20°C.
Reconstitute to a concentration ≥1 µg/ml in 4 mM sterile HCl containing ≥0.1% HSA or BSA.

Other Notes

Shibuya, H., et al. 1996. Science272, 1179.
Bottlinger, E.P., et al. 1995. Proc. Natl. Acad. Sci. USA93, 5877.
Lomo, J., et al. 1995. J. Immunol.154, 1634.
Marushige, K., and Marushige, Y. 1995. Anticancer Res.14, 2419.
Yamazaki, M., et al. 1994. Am. J. Pathol. 144, 221.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Holly B Hindman et al.
Investigative ophthalmology & visual science, 51(4), 1935-1942 (2009-11-13)
Purpose. To characterize phenotypic differences between anterior and posterior corneal keratocytes after stimulation with the profibrotic agent transforming growth factor-beta1 (TGF-beta1) in vitro. Methods. Sixteen corneas from healthy felines were obtained immediately after death. Lamellar dissection was performed to separate
Hon Wai Koon et al.
The American journal of pathology, 177(5), 2300-2309 (2010-10-05)
Substance P (SP) and the neurokinin-1 receptor (NK-1R) are involved in the development of colitis and mucosal healing after colonic inflammation. We studied whether SP modulates colonic fibrosis by using a chronic model of trinitrobenzenesulfonic acid (TNBS)-induced colitis in wild-type
Nagat Frara et al.
Connective tissue research, 59(1), 30-44 (2017-04-13)
Transforming growth factor beta 1 (TGFbeta-1) and connective tissue growth factor (CCN2) are important mediators of tissue repair and fibrosis, with CCN2 functioning as a downstream mediator of TGFβ-1. Substance P (SP) is also linked to collagen production in tenocytes.
Jeeranan Manokawinchoke et al.
Cell death & disease, 10(10), 761-761 (2019-10-09)
Mechanical force regulates periodontal ligament cell (PDL) behavior. However, different force types lead to distinct PDL responses. Here, we report that pretreatment with an intermittent compressive force (ICF), but not a continuous compressive force (CCF), promoted human PDL (hPDL) osteogenic
M Herranz-Itúrbide et al.
Redox biology, 40, 101841-101841 (2021-01-26)
Liver is a unique organ in displaying a reparative and regenerative response after acute/chronic damage or partial hepatectomy, when all the cell types must proliferate to re-establish the liver mass. The NADPH oxidase NOX4 mediates Transforming Growth Factor-beta (TGF-β) actions

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service