Select a Size
| Size/SKU | Availability | Price |
|---|
About This Item
recombinant
expressed in E. coli
Quality Segment
assay
>98% (SDS-PAGE and HPLC)
impurities
<0.1 μg/mg protein endotoxin (<1EU/μg)
NCBI accession no.
UniProt accession no.
shipped in
dry ice
storage temp.
-10 to -25°C
General description
Application
Stem Cell Research
Growth Factors & Receptors
Biochem/physiol Actions
Chicken, Mouse, Rat
Physical form
Preparation Note
Analysis Note
Disclaimer
1 of 1
This Item | |||
|---|---|---|---|
| assay >98% (SDS-PAGE and HPLC) | assay >98% (SDS-PAGE and HPLC) | assay - | assay >98% (SDS-PAGE and HPLC) |
| recombinant expressed in E. coli | recombinant expressed in E. coli | recombinant - | recombinant - |
| storage temp. -10 to -25°C | storage temp. -10 to -25°C | storage temp. -10 to -25°C | storage temp. -10 to -25°C |
| impurities <0.1 μg/mg protein endotoxin (<1EU/μg) | impurities <0.1 μg/mg protein endotoxin (<1EU/μg) | impurities <0.1 μg/mg protein endotoxin (<1 EU/μg) | impurities <0.1 μg/mg protein endotoxin (<1 EU/μg) |
| shipped in dry ice | shipped in dry ice | shipped in dry ice | shipped in dry ice |
| UniProt accession no. | UniProt accession no. | UniProt accession no. | UniProt accession no. |
Storage Class
11 - Combustible Solids
wgk
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



