Skip to Content
Merck
All Photos(1)

Key Documents

6.04717

Sigma-Aldrich

Isooctane

for chromatography

Synonym(s):

2,2,4-Trimethylpentane, Isooctane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCH2C(CH3)3
CAS Number:
Molecular Weight:
114.23
Beilstein:
1696876
MDL number:
UNSPSC Code:
12352001

grade

for chromatography

Quality Level

vapor density

3.9 (vs air)

vapor pressure

41 mmHg ( 21 °C)

autoignition temp.

745 °F

expl. lim.

6 %

refractive index

n20/D 1.391 (lit.)

bp

98-99 °C (lit.)

mp

−107 °C (lit.)

density

0.692 g/mL at 25 °C (lit.)

suitability

suitable for chromatography

SMILES string

CC(C)CC(C)(C)C

InChI

1S/C8H18/c1-7(2)6-8(3,4)5/h7H,6H2,1-5H3

InChI key

NHTMVDHEPJAVLT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Isooctaneis a volatile organic solvent used in chromatography, often as an exchange solventfor analyzing multiclass pesticide residues together with hexane and toluene.

Application

Isooctane can be used as:
  • A solvent in the liquid chromatograph-massspectrometer-computer analytical system for the separation of several polynuclear hydrocarbons.
  • One of the eluent solvents in the normal-phase analysis of lipids.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

10.4 °F - closed cup

Flash Point(C)

-12 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Evaluation of common organic solvents for gas chromatographic analysis and stability of multiclass pesticide residues
Mastovska K and Lehotay SJ
Journal of Chromatography A, 1040(2), 259-272 (2004)
Atmospheric pressure ionization mass spectrometry. Corona discharge ion source for use in a liquid chromatograph-mass spectrometer-computer analytical system
Carroll DI, et al.
Analytical Chemistry, 47(14), 2369-2373 (1975)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service