Skip to Content
Merck
All Photos(2)

Key Documents

1.04717

Supelco

Isooctane

for liquid chromatography LiChrosolv®

Synonym(s):

2,2,4-Trimethylpentane, 2,2,4-Trimethylpentane, Isobutyltrimethylmethane, iso-Octane, Isooctane

Sign Into View Organizational & Contract Pricing

Select a Size

1 L
€160.00
2.5 L
€334.00

€160.00


Estimated to ship on22 May 2025


Request a Bulk Order

Select a Size

Change View
1 L
€160.00
2.5 L
€334.00

About This Item

Linear Formula:
(CH3)2CHCH2C(CH3)3
CAS Number:
Molecular Weight:
114.23
Beilstein:
1696876
MDL number:
UNSPSC Code:
12191502
EC Index Number:
208-759-1
NACRES:
NA.03
technique(s):
HPLC: suitable
bp:
98-99 °C (lit.)
vapor pressure:
41 mmHg ( 21 °C)

€160.00


Estimated to ship on22 May 2025


Request a Bulk Order

vapor density

3.9 (vs air)

Quality Level

vapor pressure

41 mmHg ( 21 °C)

description

isocratic

product line

LiChrosolv®

Assay

≥99% (GC)

form

liquid

autoignition temp.

745 °F

potency

>2500 mg/kg LD50, oral (Rat)

expl. lim.

6 %

technique(s)

HPLC: suitable

impurities

≤0.0002 meq/g Acidity
≤0.0002 meq/g Alkalinity
≤0.01% Water

evapn. residue

≤2.0 mg/L

transmittance

210 nm, ≥50%
220 nm, ≥80%
245 nm, ≥98%

refractive index

n20/D 1.391 (lit.)

pH

7

bp

98-99 °C (lit.)

mp

−107 °C (lit.)

transition temp

flash point -12 °C

density

0.692 g/mL at 25 °C (lit.)

storage temp.

2-30°C

SMILES string

CC(C)CC(C)(C)C

InChI

1S/C8H18/c1-7(2)6-8(3,4)5/h7H,6H2,1-5H3

InChI key

NHTMVDHEPJAVLT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

With their high degree of UV transmittance, low particle count, low acidity and alkalinity and low evaporation residue level, LiChrosolv solvents are ideal for reproducible separations. Since separations are normally carried out under gradient conditions in analytical HPLC, we offer solvents in "gradient grade" as well as "isocratic grade". This enables to minimize the gradient effect of the solvent involved.

Application


  • Spectral Investigations of Fluorescence Tracers in Automotive and Aviation Fuels under Cryogenic Conditions: This research highlights the use of isooctane in studying the behavior of fuels at low temperatures, proving essential for improving fuel performance and safety in automotive and aviation industries (Koegl et al., 2024).

  • Versatile Application of TiO2@PDA Modified Filter Paper for Oily Wastewater Treatment: Demonstrates isooctane′s role in assessing the efficiency of innovative materials designed for environmental applications, specifically in the treatment of oily wastewaters, enhancing sustainability in industrial practices (Zhao et al., 2023).

  • Characterization and Pilot-Scale Application of the ZMS-2 Serine Protease with Novel Properties for the Eco-friendly Leather Processing: Utilizes isooctane in extracting and characterizing enzymes that significantly reduce the environmental impact of leather processing, marking a step forward in sustainable industrial manufacturing (Khan et al., 2023).

  • Rapid Construction of Caffeic Acid/p-Phenylenediamine Antifouling Hydrophilic Coating on a PVDF Membrane for Emulsion Separation: Investigates the role of isooctane in creating advanced coating solutions that improve the efficiency and longevity of separation processes, critical for environmental engineering (Tong et al., 2023).

Specifications

Saturation Concentration in Air: 239 g/m3 (20 °C)

Preparation Note

Product filtered through a 0.2 μm filter

Analysis Note

Purity (GC): ≥ 99.0 %
Identity (IR): conforms
Evaporation residue: ≤ 2.0 mg/l
Water: ≤ 0.01 %
Acidity: ≤ 0.0002 meq/g
Alkalinity: ≤ 0.0002 meq/g
Transmission (at 210 nm): ≥ 50 %
Transmission (at 220 nm): ≥ 80 %
Transmission (from 245 nm): ≥ 98 %
Filtered by 0.2 µm filter

Other Notes

Explore various lab safety accessories and equipment for safe handling of solvents to increase your safety level from the first usage.

Legal Information

LICHROSOLV is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

10.4 °F - closed cup

Flash Point(C)

-12 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service