Skip to Content
Merck
All Photos(1)

Key Documents

580566

Sigma-Aldrich

Tenovin-1

A cell-permeable benzoylthiourea compound that acts as a reversible inhibitor of class III HDAC sirtuins against SirT2 (IC₅₀ = ~ 10 µM).

Synonym(s):

Tenovin-1, N-(4- tert-Butylbenzoyl)-Nʹ-(4-acetylaminophenyl)-thiourea, SIRT2 Inhibitor V, N-(4-tert-Butylbenzoyl)-Nʹ-(4-acetylaminophenyl)-thiourea, SIRT2 Inhibitor V

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€74.00

€74.00


Estimated to ship on30 May 2025



Select a Size

Change View
10 MG
€74.00

About This Item

Empirical Formula (Hill Notation):
C20H23N3O2S
CAS Number:
Molecular Weight:
369.48
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€74.00


Estimated to ship on30 May 2025


Quality Level

Assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 5 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

S=C(Nc2ccc(cc2)NC(=O)C)NC(=O)c1ccc(cc1)C(C)(C)C

InChI

1S/C20H23N3O2S/c1-13(24)21-16-9-11-17(12-10-16)22-19(26)23-18(25)14-5-7-15(8-6-14)20(2,3)4/h5-12H,1-4H3,(H,21,24)(H2,22,23,25,26)

InChI key

WOWJIWFCOPZFGV-UHFFFAOYSA-N

General description

A cell-permeable benzoylthiourea compound that acts as a reversible inhibitor of class III HDAC sirtuins against SirT2 (IC50 = ~ 10 µM). Based on studies using the more soluble analog Tenovin-6, Tenovin-1 is expected not to compete with substrate binding and display lower potency against SirT1/3 (IC50 = 21/67 µM using Tenovin-6). Tenovin-1 (10 µM) up-regulates cellular p53 protein, but not mRNA, level in MCF-7 (≥6-fold in 6 h), presumably by blocking mdm2-mediated p53 degradation. Two day Tenovin-1 treatment (10 µM) results in selective killing of cancer cells with functional p53, but not p53-lacking cancer cells or normal human dermal fibroblasts. Despite its low aqueous solubility, administration of Tenovin-1 suspension is shown to effectively reduce BL2 and ARN8 xenographs in mice in vivo (≥50% reduction with 92 mg/kg, daily, i.p.).

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Lain, S., et al. 2008. Cancer Cell13, 454.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service