Skip to Content
Merck
All Photos(1)

Key Documents

PZ0336

Sigma-Aldrich

TMI-1

≥98% (HPLC)

Synonym(s):

4-[[4-(2-Butynyloxy)phenyl]sulfonyl]-N-hydroxy-2,2-dimethyl-(3S)thiomorpholinecarboxamide, TMI 1, WAY-171318

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€102.00
25 MG
€412.00

€102.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€102.00
25 MG
€412.00

About This Item

Empirical Formula (Hill Notation):
C17H22N2O5S2
CAS Number:
Molecular Weight:
398.50
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

€102.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 25 mg/mL, clear

storage temp.

room temp

SMILES string

O=S(N1[C@@H](C(NO)=O)C(C)(C)SCC1)(C2=CC=C(OCC#CC)C=C2)=O

InChI

1S/C17H22N2O5S2/c1-4-5-11-24-13-6-8-14(9-7-13)26(22,23)19-10-12-25-17(2,3)15(19)16(20)18-21/h6-9,15,21H,10-12H2,1-3H3,(H,18,20)/t15-/m0/s1

InChI key

CVZIHNYAZLXRRS-HNNXBMFYSA-N

Biochem/physiol Actions

TMI-1 (WAY-171318) is potent inhibitor of disintegrin metalloenzyme 17 (ADAM17) also known as the tumor necrosis factor-α (TNF-α) converting enzyme (TACE), and matrix metalloprotease (MMP). TMI-1 potently inhibits LPS-induced TNF-α secretion in human primary monocytes, and human whole blood. TMI-1 selectively induces caspase-dependent apoptosis in triple negative (TN) and ERBB2-overexpressing breast tumor cell lines.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service