Skip to Content
Merck
All Photos(1)

Key Documents

569398

Sigma-Aldrich

Stearic Acid

Synonym(s):

Stearic Acid, Octadecanoic Acid

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
€157.00
100 G
€441.00

€157.00


Estimated to ship on18 May 2025


Request a Bulk Order

Select a Size

Change View
25 G
€157.00
100 G
€441.00

About This Item

Empirical Formula (Hill Notation):
C18H36O2
CAS Number:
Molecular Weight:
284.48
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.77

€157.00


Estimated to ship on18 May 2025


Request a Bulk Order

description

Merck USA index - 14, 8804

Quality Level

Assay

≥99% (GC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

impurities

≤0.5% saponifiable substance, as methyl ester

color

white

solubility

ethanol: soluble

shipped in

ambient

storage temp.

10-30°C

SMILES string

[O-]C(=O)CCCCCCCCCCCCCCCCC.[H+]

InChI

1S/C18H36O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20/h2-17H2,1H3,(H,19,20)

InChI key

QIQXTHQIDYTFRH-UHFFFAOYSA-N

General description

Stearic Acid is a common fatty acid that isfound as a glycerol ester in most plant and animal fats. It exists athigher concentrations in animal fats than plant fats. Stearic acid is found asa triglyceride in cocoa butter and shea butter. It is used to enrich foodinstead of palmitic acid and trans-fat in the food industry. Stearic acid isapplicable in the production of soaps, detergents, shampoos, shaving creams, andcosmetics.[1]

Application

Stearic acid has been used as a light chainfatty acid (LCFA) to study its effects on the dysfunction of tumor-derived exosomelipid-induced dendritic cells.[2]

Biochem/physiol Actions

Cell permeable: no
Product does not compete with ATP.
Reversible: no

Warning

Toxicity: Standard Handling (A)

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

392.0 °F - Cleveland open cup

Flash Point(C)

200 °C - Cleveland open cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Diego Cuartas-Marulanda et al.
Polymers, 14(23) (2022-12-12)
Magnesium (Mg) alloys have great potential in biomedical applications due to their incomparable properties regarding other metals, such as stainless steels, Co-Cr alloys, and titanium (Ti) alloys. However, when Mg engages with body fluids, its degradation rate increases, inhibiting the
Xiaozhe Yin et al.
Cell reports, 33(3), 108278-108278 (2020-10-22)
Dendritic cells (DCs) orchestrate the initiation, programming, and regulation of anti-tumor immune responses. Emerging evidence indicates that the tumor microenvironment (TME) induces immune dysfunctional tumor-infiltrating DCs (TIDCs), characterized with both increased intracellular lipid content and mitochondrial respiration. The underlying mechanism

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service