Skip to Content
Merck
All Photos(1)

Key Documents

8.00673

Sigma-Aldrich

Stearic acid

for synthesis

Synonym(s):

Stearic acid

Sign Into View Organizational & Contract Pricing

Select a Size

500 G
€37.70
1 KG
€43.30
50 KG
€749.00

€37.70


Estimated to ship on23 March 2025


Request a Bulk Order

Select a Size

Change View
500 G
€37.70
1 KG
€43.30
50 KG
€749.00

About This Item

Linear Formula:
CH3(CH2)16COOH
CAS Number:
Molecular Weight:
284.48
MDL number:
UNSPSC Code:
12352106
EC Index Number:
200-313-4
NACRES:
NA.22

€37.70


Estimated to ship on23 March 2025


Request a Bulk Order

Quality Level

Assay

≥97% (GC)

form

solid

autoignition temp.

395 °C

potency

>5000 mg/kg LD50, oral (Rat)
>5000 mg/kg LD50, skin (Rabbit)

bp

370 °C/1013 hPa

mp

68-70 °C

transition temp

flash point 196 °C

solubility

water: insoluble at 20 °C

bulk density

400‑500 kg/m3

storage temp.

2-30°C

SMILES string

[O-]C(=O)CCCCCCCCCCCCCCCCC.[H+]

InChI

1S/C18H36O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20/h2-17H2,1H3,(H,19,20)

InChI key

QIQXTHQIDYTFRH-UHFFFAOYSA-N

Related Categories

Application


  • Green Strong Cornstalk Rind-Based Cellulose-PVA Aerogel for Oil Adsorption and Thermal Insulation: This study highlights a novel aerogel incorporating stearic acid for enhanced oil adsorption and thermal insulation properties (X Yi et al., 2024).

  • Preparation and characterization of self-aggregating micelles from stearic acid-modified burdock root polysaccharide: Focuses on the development of micelles using stearic acid, exploring their potential in food science and technology (C Zhang et al., 2024).

  • A prototype system for the removal of 137Cs from liquid radioactive waste using reverse osmosis membrane: Discusses the use of a stearic acid-modified membrane for improving radioactive waste treatment (M Shaltout et al., 2024).


  • Compatibility study of mirtazapine with several excipients used in pharmaceutical dosage forms employing thermal and non-thermal methods: Investigates interactions between stearic acid and pharmaceutical excipients, indicating possible concerns for drug formulation (D Circioban et al., 2024).

Analysis Note

Assay (GC, area%): ≥ 97.0 % (a/a)
Melting range (lower value): ≥ 68 °C
Melting range (upper value): ≤ 70 °C
Identity (IR): passes test

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

392.0 °F - Cleveland open cup

Flash Point(C)

200 °C - Cleveland open cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service