Skip to Content
Merck
All Photos(1)

Key Documents

567549

Millipore

Sodium Phosphate, Monobasic, Monohydrate, Molecular Biology Grade

Useful in conjunction with Sodium Phosphate, Dibasic in the preparation of biological buffers.

Synonym(s):

Sodium phosphate monobasic monohydrate, NaH₂PO₄, Monosodium phosphate, Sodium dihydrogen phosphate monohydrate

Sign Into View Organizational & Contract Pricing

Select a Size

250 G
€84.20
1 KG
€296.00

€84.20


Estimated to ship on29 May 2025


Request a Bulk Order

Select a Size

Change View
250 G
€84.20
1 KG
€296.00

About This Item

Linear Formula:
NaH2PO4 · H2O
CAS Number:
Molecular Weight:
137.99
MDL number:
UNSPSC Code:
12352302
NACRES:
NA.25

€84.20


Estimated to ship on29 May 2025


Request a Bulk Order

grade

Molecular Biology

Quality Level

description

Merck USA index - 14, 8660

Assay

≥99% (titration)

form

crystalline solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

color

white

pKa (20 °C)

7.21

solubility

water: 14 mg/mL

cation traces

heavy metals: ≤0.001%
heavy metals (as Pb): ≤0.001%

absorption

≤0.01 at 260 nm
≤0.01 at 280 nm

foreign activity

DNases, proteases, RNases, none detected

shipped in

ambient

storage temp.

10-30°C

SMILES string

[Na+].[H]O[H].OP(O)([O-])=O

InChI

1S/Na.H3O4P.H2O/c;1-5(2,3)4;/h;(H3,1,2,3,4);1H2/q+1;;/p-1

InChI key

BBMHARZCALWXSL-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Useful in conjuction with sodium phosphate, dibasic (Cat. No. 567550) in the preparation of biological buffers(absorbance: ≤0.01 at 260 nm and 280 nm).
Useful in conjunction with Sodium Phosphate, Dibasic (Cat. No. 567550 or 567547) in the preparation of biological buffers.

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, store at room temperature. Stock solutions are stable for up to 6 months at room temperature.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service