Skip to Content
Merck
All Photos(1)

Key Documents

567545

Millipore

Sodium Phosphate, Monobasic

Sodium Phosphate, Monobasic

ACS Reagent Grade. Useful in conjunction with Sodium Phosphate, Dibasic in the preparation of biological buffers.

Synonym(s):

Sodium phosphate monobasic monohydrate, NaH₂PO₄, Monosodium phosphate, Sodium dihydrogen phosphate monohydrate

Sign Into View Organizational & Contract Pricing

Select a Size

500 G
€91.50
1 KG
€143.00

€91.50


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
500 G
€91.50
1 KG
€143.00

About This Item

Linear Formula:
NaH2PO4 · H2O
CAS Number:
Molecular Weight:
137.99
MDL number:
UNSPSC Code:
12352302
NACRES:
NA.25

€91.50


Estimated to ship on28 May 2025


Request a Bulk Order

grade

ACS reagent

Quality Level

description

Merck USA index - 14, 8660

Assay

≥98% (by assay)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

pKa (20 °C)

7.21

solubility

water: soluble

cation traces

heavy metals: ≤0.001%

shipped in

ambient

storage temp.

10-30°C

SMILES string

[Na+].[H]O[H].OP(O)([O-])=O

InChI

1S/Na.H3O4P.H2O/c;1-5(2,3)4;/h;(H3,1,2,3,4);1H2/q+1;;/p-1

InChI key

BBMHARZCALWXSL-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

ACS Reagent Grade. Useful in conjunction with Sodium Phosphate, Dibasic (Cat. No. 567550 or 567547) in the preparation of biological buffers. Has a pH of 4.1-4.5.

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, store at room temperature. Stock solutions are stable for several months at room temperature.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

Active Filters

  1. 1 Ratings-Only Review

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service