Skip to Content
Merck
All Photos(1)

Key Documents

539578

Sigma-Aldrich

PKG, Iα, Bovine Lung

Synonym(s):

cGMP-Dependent Protein Kinase Catalytic Subunit, PKG Catalytic Subunit, Protein Kinase G Iα, cGMP-Dependent Protein Kinase Catalytic Subunit, PKG Catalytic Subunit, Protein Kinase G Iα

Sign Into View Organizational & Contract Pricing

Select a Size

4 μG
€467.00

€467.00


Estimated to ship on11 May 2025



Select a Size

Change View
4 μG
€467.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.42

€467.00


Estimated to ship on11 May 2025


form

liquid

Quality Level

specific activity

≥1300 units/μg protein

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

shipped in

wet ice

storage temp.

−70°C

General description

Note: 4µg = 6000 units.
Native PKGIα purified from bovine lung. PKG is involved in regulation of smooth muscle relaxation, platelet function, and cell division. PKG is activated by binding of cGMP to regulatory subunits within the holoenzyme, but in contrast to PKA, the activation of PKG does not invlove physical dissociation of catalytic from the regulatory domains. The activated enzyme phosphorylates target proteins at serine and/or threonine residues that are found within the recognition motif, Arg-Lys-Arg-Ser*-Arg-Ala-Glu.

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Unit Definition

One unit is defined as the amount of enzyme required to incorporote 1 pmol phosphate into the synthetic heptapeptide substrate, RKRSRAE, in 1 min at 30°C.

Physical form

In 150 mM NaCl, 25 mM β-mercaptoethanol, 10 mM K₂HPO₄, 1 mM EDTA, 12% sucrose, pH 6.8.

Reconstitution

Following initial thaw, aliquot and freeze (-70°C).

Other Notes

Brophy, M.C., et al. 2006. J. Vasc. Res.39, 95.
Colbran J.L., et al. 1992. J. Biol. Chem.267, 9589.
Corbin, J.D. and Doskeland, S.O. 1983. J Biol Chem258, 11391.
Witt, J.J. and Roskoski, R., Jr, 1975. Anal Biochem66, 253.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service