Skip to Content
Merck
All Photos(1)

Key Documents

5.33683

Sigma-Aldrich

PMX53

≥98% (HPLC), solid, C5aR antagonist, Calbiochem®

Synonym(s):

C5aR Antagonist, PMX53, Ac-(cyclo-2,6)-F-[OPdChaWR], C5aR Inhibitor, PMX53, C5a Receptor Inhibitor, PMX53, CD88 Inhibitor, PMX53, MrgX2 Agonist, PMX53

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€238.00

€238.00


Estimated to ship on24 May 2025


Request a Bulk Order

Select a Size

Change View
10 MG
€238.00

About This Item

Empirical Formula (Hill Notation):
C47H65N11O7 · xC2HF3O2
CAS Number:
Molecular Weight:
896.09 (free base basis)
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€238.00


Estimated to ship on24 May 2025


Request a Bulk Order

Product Name

C5aR Antagonist, PMX53,

Assay

≥98% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 50 mg/mL

storage temp.

2-8°C

General description

A cell-permeable, orally available, and metabolically stable macrocyclic hexapeptidomimetic compound that acts as a non-competitive antagonist of complement C5a receptor (C5aR; CD88; IC50 = 240 nM) and as a low-affinity agonist for MrgX2, a GPCR, in human mast cells Shown to specifically bind to C5aR1 and not to CaR2 and C3aR. Displays a broad range of anti-inflammatory effects, both in vitro and in vivo. Shown to inhibit C5a-induced neutrophil myeloperoxidase release (IC50 = 22 nM) and chemotaxis (IC50 = 75 nM). Preincubation with PMX-53 (~10 nM) blocks C5a-induced Ca2+ responses in HMC-1 and RBL-2H3 cells, but does not affect C3a responses. Suppresses the growth of 4T1 mammary carcinoma tumor xenograft in mice (1 mg/kg/every 2-3 days, s.c.). Reported to diminish C5a-mediated increase in pluripotency in human embryonic stem cells in the absence of FGF2.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
C5aR
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Sequence

Ac-Phe-[Orn-Pro-dCha-Trp-Arg]

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Lillegard, K., et al. 2014. JPET.351,344.
Vadrevu, S. K., et al. 2014. Cancer Res.74, 3454.
Liang, S., et al. 2011. J. Immun.186, 869.
Woodruff, T., et al. 2005. JPET.314, 811.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service