Skip to Content
Merck
All Photos(1)

Key Documents

525145

Sigma-Aldrich

sPLA2-IIA Inhibitor I

The sPLA₂-IIA Inhibitor I controls the biological activity of sPLA₂-IIA. This small molecule/inhibitor is primarily used for Cell Signaling applications.

Synonym(s):

sPLA2-IIA Inhibitor I, c(2NapA)LS(2NapA)R, TFA

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€177.00

€177.00


Estimated to ship on25 May 2025



Select a Size

Change View
1 MG
€177.00

About This Item

Empirical Formula (Hill Notation):
C41H50N8O6 · xC2HF3O2
Molecular Weight:
750.89 (free base basis)
UNSPSC Code:
41106300
NACRES:
NA.77

€177.00


Estimated to ship on25 May 2025


Quality Level

Assay

≥95% (HPLC)

form

lyophilized solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

color

white

solubility

DMSO: 5 mg/mL

shipped in

ambient

storage temp.

−20°C

General description

A highly hydrophobic cyclic pentapeptide that selectively binds and acts as a potent inhibitor of sPLA2-IIA (human type IIA secreted phospholipase A2; IC50 = 12.8 μM). Shown to effectively block sPLA2-IIA-induced PGE2 production at 100 nM in human rheumatoid synoviocytes and is non-toxic at doses up to 10 μM. Does not have any significant effect on the activities of porcine sPLA2-IB, Naja naja sPLA2-IB, or Crotalus durissus sPLA2-IIA even at 10 μM concentration.

Biochem/physiol Actions

Cell permeable: no
Primary Target
sPLA2-IIA (human type IIA secreted phospholipase A2
Product does not compete with ATP.
Reversible: no
Target IC50: 12.8 µM against sPLA2-IIA (human type IIA secreted phospholipase A2

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Sequence

cyclic(2-NaphthylAla-Leu-Ser-2-NaphthylAla-Arg)•TFA

Other Notes

Church, W.B., et al. 2001. J. Biol. Chem.276, 33156.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Julien Pothlichet et al.
Frontiers in immunology, 13, 824746-824746 (2022-04-09)
The origin of the impaired CD4 T-cell response and immunodeficiency of HIV-infected patients is still only partially understood. We recently demonstrated that PLA2G1B phospholipase synergizes with the HIV gp41 envelope protein in HIV viremic plasma to induce large abnormal membrane

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service