Skip to Content
Merck
All Photos(1)

Key Documents

5.33575

Sigma-Aldrich

Rigosertib

Synonym(s):

Rigosertib, Sodium ( E)-2-(2-methoxy-5-((2,4,6-trimethoxystyrylsulfonyl)methyl)phenylamino)acetate, ON01910, Na, Estybon

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€185.00

€185.00


Estimated to ship on16 May 2025


Request a Bulk Order

Select a Size

Change View
10 MG
€185.00

About This Item

Empirical Formula (Hill Notation):
C21H24NNaO8S
CAS Number:
Molecular Weight:
473.47
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€185.00


Estimated to ship on16 May 2025


Request a Bulk Order

Assay

≥98% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 50 mg/mL

storage temp.

2-8°C

SMILES string

[Na+].[S](=O)(=O)(Cc2cc(c(cc2)OC)NCC(=O)[O-])\C=C\c1c(cc(cc1OC)OC)OC

InChI

1S/C21H25NO8S.Na/c1-27-15-10-19(29-3)16(20(11-15)30-4)7-8-31(25,26)13-14-5-6-18(28-2)17(9-14)22-12-21(23)24;/h5-11,22H,12-13H2,1-4H3,(H,23,24);/q;+1/p-1/b8-7+;

InChI key

VLQLUZFVFXYXQE-USRGLUTNSA-M

General description

A cell-permeable, nontoxic benzylstyrylsulfone based compound with anti-tumor and radiosensitizing properties. Potently suppresses PI 3-K/Akt/mTOR signaling pathway and activates Cdc25C phosphatases. Acts as an allosteric, substrate-competitive, and reversible inhibitor of multiple protein kinases (IC50 = 32, 42, 182, 18 and 155 nM against Bcr-Abl, Flt1, Fyn, PDGFR, and Src, respectively). Induces DNA damage and blocks microtubule formation. Also shown to cause mitotic arrest and apoptosis in multiple types of cancer cells (GI50 = 30 - 200 nM) and blocks cell cycle at G2/M phase. Effective against several multi-drug resistance tumor cells (IC50 = 50-100 nM). Suppresses the growth of BT20 tumor xenografts in mice and improves their survival rate (200 mg/kg, i.v. Q2D X 20 schedule). However, it displays limited oral bioavailability.
Rigosertib, CAS 592542-60-4, is a cell-permeable, potent suppressor of PI 3-K/Akt/mTOR signaling. Allosterically inhibits several key protein kinases that are over active in many cancer cells.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
PI 3-K/Akt/mTOR signaling
Target IC50: 32, 42, 182, 18 and 155 nM against Bcr-Abl, Flt1, Fyn, PDGFR, and Src, respectively

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Lu, T., et al. 2015. Mol. Pharmacol.87, 18.
Ramana Reddy, M.V., et al. 2011. J. Med. Chem.54, 6254.
Gumireddy, K., et al. 2005. Cancer Cell7, 275.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service