Skip to Content
Merck
All Photos(1)

Key Documents

5.32177

Sigma-Aldrich

Tdp1 Inhibitor, CD00509

Synonym(s):

Tdp1 Inhibitor, CD00509, Tyrosyl-DNA phosphodiesterase 1 Inhibitor

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
€198.00

€198.00


Estimated to ship on31 May 2025


Request a Bulk Order

Select a Size

Change View
25 MG
€198.00

About This Item

Empirical Formula (Hill Notation):
C9H6N2O3S
CAS Number:
Molecular Weight:
222.22
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€198.00


Estimated to ship on31 May 2025


Request a Bulk Order

Assay

≥97% (HPLC)

Quality Level

form

powder

potency

710 nM IC50

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow

solubility

DMSO: soluble

storage temp.

2-8°C

SMILES string

S=C1NC(=O)C(=Cc2[o]ccc2)C(=O)N1

InChI

1S/C9H6N2O3S/c12-7-6(4-5-2-1-3-14-5)8(13)11-9(15)10-7/h1-4H,(H2,10,11,12,13,15)

InChI key

VQRARDATQWRHGF-UHFFFAOYSA-N

General description

A cell permeable alkylidene barbiturate analog that acts as a potent and specific inhibitor of tyrosyl-DNA phosphodiesterase 1 (Tdp1; IC50 = 710 nM) and increases the number of g-H2AX foci in MCF-7 cells. Shown to sensitize MCF-7 cells to camptothecin (CPT), increase DNA breaks, and reduce cell proliferation. However, it does not affect the viability of normal mammary epithelial cells up to 10 µM levels. Synergistically reduces cell proliferation and induces apoptosis in MCF-7 cells when combined with Rucaparib, a PARP-1 inhibitor.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Tdp1

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Dean, R.A., et al. 2014. J. Biolmol. Screen.19, 1372.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service