Skip to Content
Merck
All Photos(1)

Key Documents

5310-OP

Millipore

HEPES

>= 99%, Free Acid, fine powder, OmniPur® Grade

Synonym(s):

HEPES, N-2-Hydroxyethylpiperazine-N′-2-ethanesulfonic acid, 4-(2-Hydroxyethyl)piperazine-1-ethanesulfonic acid, N-(2-Hydroxyethyl)piperazine-N′-(2-ethanesulfonic acid)

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
Inquire
500 G
Inquire
1 KG
€592.00
5 KG
Inquire


Estimated to ship on22 May 2025


Request a Bulk Order

Select a Size

Change View
100 G
Inquire
500 G
Inquire
1 KG
€592.00
5 KG
Inquire

About This Item

Empirical Formula (Hill Notation):
C8H18N2O4S
CAS Number:
Molecular Weight:
238.30
Beilstein:
883043
MDL number:
UNSPSC Code:
12352207
NACRES:
NA.21


Estimated to ship on22 May 2025


Request a Bulk Order

Product Name

HEPES, OmniPur® Grade, >= 99%, Free Acid

Quality Level

form

fine powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

useful pH range

6.8-8.2

pKa (25 °C)

7.5

shipped in

ambient

storage temp.

10-30°C

SMILES string

OCCN1CCN(CC1)CCS(O)(=O)=O

InChI

1S/C8H18N2O4S/c11-7-5-9-1-3-10(4-2-9)6-8-15(12,13)14/h11H,1-8H2,(H,12,13,14)

InChI key

JKMHFZQWWAIEOD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description



Molecular Biology grade

A260 (0.1 M, water): ≤0.05 AU
A280 (0.1 M, water): ≤0.05 AU
Assay (dried basis): ≥99
DNase: None detected
Heavy metals (as Pb): ≤0.001%
Phosphatase: None detected
pKa at 25°C: 6.80-8.20
Protease: None detected
RNase: None detected





Intended for laboratory and manufacturing use only. Not for drug, food, or household use.

Application

<ul>
<li><strong>Phenothiazine appended thiophene derivative: a trilateral approach to copper ion detection in living cells and aqueous samples.</strong>: This study explores the use of HEPES buffer in the detection of copper ions, showcasing its utility in maintaining the stability of the sensor in aqueous solutions and live cell environments. The findings indicate that HEPES buffer plays a crucial role in the accuracy and efficiency of ion detection assays, which is essential for biochemists working with metal ion sensors (Ilakiyalakshmi et al., 2024).</li>
<li><strong>Evaluation of acute intravenous toxicity of HEPES: Is Good′s buffer good and safe enough for clinical utilization in nuclear medicine </strong>: This research assesses the safety of HEPES buffer for intravenous use, specifically its acute toxicity. The results are vital for biochemists in nuclear medicine, indicating that HEPES is a safe and effective buffering agent for clinical applications (Guleria et al., 2024).</li>
<li><strong>Nanoscale engineering of gold nanostars for enhanced photoacoustic imaging.</strong>: The study highlights the use of HEPES buffer in the synthesis of gold nanostars, which significantly improves photoacoustic imaging. This demonstrates the buffer′s importance in nanobiotechnology, particularly in enhancing the stability and functionality of nanomaterials used in imaging techniques (Zhang et al., 2024).</li>
<li><strong>Influx of zwitterionic buffer after intracytoplasmic sperm injection (ICSI) membrane piercing alters the transcriptome of human oocytes.</strong>: This article discusses how HEPES buffer influences the transcriptome of human oocytes post-ICSI, highlighting its role in reproductive biology and assisted reproductive technologies. The findings are crucial for biochemists focusing on fertility treatments and genetic studies (Mendola et al., 2024).</li>
<li><strong>One Method to Label Them All": A Single Fully Automated Protocol for GMP-Compliant 68Ga Radiolabeling of PSMA-11, Transposable to PSMA-I and PSMA-617.</strong>: This paper outlines a protocol using HEPES buffer for radiolabeling in nuclear medicine, emphasizing its application in producing radiopharmaceuticals. The research supports the buffer′s critical role in maintaining the quality and compliance of radiolabeling processes (Fouillet et al., 2024).</li>
</ul>

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
OMNIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Articles

Determination of free thyroid hormones (T3 & T4) in serum by LC-MS/MS following a streamlined 1-hour BioSPME sample preparation.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service