Skip to Content
Merck
All Photos(1)

Key Documents

391338

Millipore

HEPES

≥99% dry basis (titration), crystalline powder, amniosulfonic acid buffer

Synonym(s):

HEPES, N-2-Hydroxyethylpiperazine-Nʹ-2-ethanesulfonic Acid, 4-(2-Hydroxyethyl)piperazine-1-ethanesulfonic acid, N-(2-Hydroxyethyl)piperazine-N′-(2-ethanesulfonic acid)

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
€80.60
100 G
Inquire
500 G
€452.00
1 KG
Inquire
5 KG
Inquire

€80.60


Estimated to ship on24 May 2025


Request a Bulk Order

Select a Size

Change View
25 G
€80.60
100 G
Inquire
500 G
€452.00
1 KG
Inquire
5 KG
Inquire

About This Item

Empirical Formula (Hill Notation):
C8H18N2O4S
CAS Number:
Molecular Weight:
238.30
Beilstein:
883043
MDL number:
UNSPSC Code:
12352106
NACRES:
NA.25

€80.60


Estimated to ship on24 May 2025


Request a Bulk Order

Product Name

HEPES, ULTROL® Grade, ≥99% dry basis (titration), zwitterionic N-substituted aminosulfonic acid buffer

description

Merck USA index - 14, 4654

Quality Level

Assay

≥99% dry basis (titration)

form

crystalline powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

useful pH range

6.8-8.2

pKa (25 °C)

7.5

solubility

water: 1 M

anion traces

chloride (Cl-): ≤0.01%
sulfate (SO42-): ≤0.05%

cation traces

Na: ≤0.1%
heavy metals: ≤1 ppm

shipped in

ambient

storage temp.

15-25°C

SMILES string

OCCN1CCN(CC1)CCS(O)(=O)=O

InChI

1S/C8H18N2O4S/c11-7-5-9-1-3-10(4-2-9)6-8-15(12,13)14/h11H,1-8H2,(H,12,13,14)

InChI key

JKMHFZQWWAIEOD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

A zwitterionic N-substituted aminosulfonic acid buffer. HEPES has its maximum buffering capacity very near the pH of mammalian tissue fluids (pKa of 7.31 at 37°C). Exhibits no metal binding and has a small temperature dependent ΔpKa of -0.014/°C.
Absorbance (1.0 M, H2O, 260 nm):≤0.05.

Absorbance (1.0 M, H2O, 280 nm): ≤0.04

Warning

Toxicity: Irritant (B)

Reconstitution

Following reconstitution, store at room temperature (20°C). Stock solutions are stable for up to 6 months at 20°C.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
ULTROL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service