Skip to Content
Merck
All Photos(1)

Documents

517310

Sigma-Aldrich

Anti-P-Glycoprotein Mouse mAb (C219)

liquid, clone C219, Calbiochem®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

C219, monoclonal

form

liquid

contains

≤0.1% sodium azide as preservative

species reactivity

rat, hamster, human, mouse

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

isotype

IgG1

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... ABCB1(5243)

General description

Purified mouse monoclonal antibody. Recognizes the ~170 kDa MDR1 and MDR3 isoforms of P-glycoprotein.
Recognizes the ~170 kDa MDR1 and MDR3 isoforms of P-glycoprotein.
This Anti-P-Glycoprotein Mouse mAb (C219) is validated for use in FC, Frozen Sections, Immunoblotting, ICC, IP, Paraffin Sections for the detection of P-Glycoprotein.

Immunogen

Hamster and Human
SDS-solubilized plasma membranes of a multidrug resistant (MDR) Chinese hamster ovary (CHO) cell line and a multidrug resistant human cell line

Application

Flow Cytometry (5-10 µg/1 x 10⁶ cells)

Frozen Sections (5-10 µg/ml)

Immunoblotting (1-10 µg/ml, see application references, see comments)

Immunocytochemistry (see comments)

Immunoprecipitation (see comments)

Paraffin Sections (1-10 µg/ml)

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Physical form

In PBS, 1% BSA.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C).

Analysis Note

Positive Control
Cell lines or frozen tissue, such as liver or colon

Other Notes

Also recognizes ~150 kDa sister P-glycoprotein (SPGP). Frozen sections should be fixed in acetone for 10 min at -20°C. Formalin-fixed, paraffin tissue sections should be deparaffinized and rehydrated by conventional means. Boiling samples may reduce visibility of the protein by immunoblotting (see application references). This antibody is also reported to work for immunocytochemistry and immunoprecipitation. A ~200 kDa protein that migrates in the same position as myosin is also seen by immunoblotting. Variables associated with assay conditions will dictate the proper working dilutions.
Beck, W.T., et al. 1996. Cancer Res. 56, 3010.
Toth, K., et al. 1992. Am. J. Pathol. 140, 1009.
Schinkel, A.H., et al. 1991. Cancer Res. 51, 2628.
Chan, H.S.L., et al. 1988. Lab Invest. 59, 870.

Legal Information

ImmunoblottingLee, G., et al. 2001. J. Pharmacol. Exp. Ther.299, 204.
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Linlin Su et al.
The international journal of biochemistry & cell biology, 41(12), 2578-2587 (2009-09-02)
Throughout spermatogenesis, leptotene spermatocytes traverse the blood-testis barrier (BTB) to enter the adluminal compartment of the seminiferous epithelium for continued development. At the same time, the integrity of the BTB, which is constituted by co-existing tight junctions (TJ), basal ectoplasmic
Yan Chen et al.
PloS one, 10(10), e0140918-e0140918 (2015-10-21)
The objective of the present study was to investigate the role of high-mobility group box-1 (HMGB1) in the seizure-induced P-glycoprotein (P-gp) overexpression and the underlying mechanism. Kainic acid (KA)-induced mouse seizure model was used for in vivo experiments. Male C57BL/6
Rose L Szabady et al.
The Journal of clinical investigation, 128(9), 4044-4056 (2018-08-14)
Neutrophil influx into the intestinal lumen is a critical response to infectious agents, but is also associated with severe intestinal damage observed in idiopathic inflammatory bowel disease. The chemoattractant hepoxilin A3, an eicosanoid secreted from intestinal epithelial cells by the
Wanghui Jing et al.
The Journal of pharmacology and experimental therapeutics, 366(2), 332-340 (2018-06-13)
Downregulation of P-glycoprotein (P-gp) is implicated in the pathophysiology of inflammatory bowel disease (IBD). Berberine, a principal isoquinoline alkaloid extracted from Berberis species, has been reported to exhibit therapeutic potential in IBD. In this study, we used a dextran sulfate
Linlin Su et al.
The Journal of endocrinology, 209(3), 337-351 (2011-04-08)
The blood--testis barrier (BTB) creates an immunological barrier that segregates the seminiferous epithelium into the basal and apical compartment. Thus, meiosis I/II and post-meiotic germ cell development take place in a specialized microenvironment in the apical compartment behind the BTB

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service