Skip to Content
Merck
All Photos(1)

Documents

SML2656

Sigma-Aldrich

TSQ

≥98% (HPLC)

Synonym(s):

6-Methoxy-8-p-toluenesulfonamido-quinoline, N-(6-Methoxy-8-quinolinyl)-4-methylbenzenesulfonamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H16N2O3S
CAS Number:
Molecular Weight:
328.39
MDL number:
UNSPSC Code:
12171500
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=S(C1=CC=C(C)C=C1)(NC2=C3C(C=CC=N3)=CC(OC)=C2)=O

InChI

1S/C17H16N2O3S/c1-12-5-7-15(8-6-12)23(20,21)19-16-11-14(22-2)10-13-4-3-9-18-17(13)16/h3-11,19H,1-2H3

InChI key

HKRNYOZJJMFDBV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Zn+ selective probe in the presence of Mg+ and Ca+.

Biochem/physiol Actions

TSQ (6-methoxy-8-p-toluenesulfonamido-quinoline) is a cell penetrant and specific fluorescent zinc sensor that preferentially accumulates in islet cells in a zinc content dependent manner. TSQ is commonly used to assess isolated islet purity.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jeffrey W Meeusen et al.
Inorganic chemistry, 50(16), 7563-7573 (2011-07-22)
Zn(2+) is a necessary cofactor for thousands of mammalian proteins. Research has suggested that transient fluxes of cellular Zn(2+) are also involved in processes such as apoptosis. Observations of Zn(2+) trafficking have been collected using Zn(2+) responsive fluorescent dyes. A
Mohammad R Karim et al.
Metallomics : integrated biometal science, 9(4), 391-401 (2016-12-06)
Nitric oxide (NO) is both an important regulatory molecule in biological systems and a toxic xenobiotic. Its oxidation products react with sulfhydryl groups and either nitrosylate or oxidize them. The aerobic reaction of NO supplied by diethylamine NONOate (DEA-NO) with
Timothy M Horton et al.
Cell chemical biology, 26(2), 213-222 (2018-12-12)
Diabetes is a hyperglycemic condition characterized by pancreatic β-cell dysfunction and depletion. Whereas methods for monitoring β-cell function in vivo exist, methods to deliver therapeutics to β cells are lacking. We leveraged the rare ability of β cells to concentrate zinc to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service