Skip to Content
Merck
All Photos(1)

Key Documents

516106

Millipore

Penicillin/Streptomycin, 100X, Tissue Culture Grade

A mixture of penicillin and streptomycin, supplied as 100X concentrated. Sterilized by γ-irradiation.

Synonym(s):

Penicllin G Sodium Salt, Streptomycin Sulfate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

shipped in

ambient

storage temp.

2-8°C

General description

A mixture of penicillin and streptomycin, supplied as 100X concentrated. Sterilized by γ-irradiation. Spectrum: Gram-positive and Gram-negative bacteria.
A mixture of penicillin and streptomycin, supplied as 100X concentrated. Sterilized by γ-irradiation. Spectrum: Gram-positive and Gram-negative bacteria. When reconstituted in 20 ml of sterile deionized water contains 10,000 units/ml Penicillin G and 10 mg/ml. Streptomycin Sulfate.

Warning

Toxicity: Harmful (C)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C) for long-term storage or refrigerate (4°C) for short-term storage. Stock solutions are stable for up to 6 months at -20°C or for up to 2 weeks at 4°C.
Reconstitute with 20 ml sterile, dH₂O to obtain a solution containing 10,000 units/ml penicillin G and 10 mg/ml streptomycin sulfate.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Repr. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service