Skip to Content
Merck
All Photos(1)

Key Documents

5.09374

Sigma-Aldrich

L524-0366

≥98% (HPLC), powder, Fn14 antagonist, Calbiochem®

Synonym(s):

Fn14 Antagonist, L524-0366, Fibroblast Growth Factor-inducible 14 Antagonist; L5240366, THD Receptor Antagonist; TNF-like Weak Inducer of Apoptosis Receptor Antagonist, TWEAK Receptor Antagonist; TWEAK-Fn14 Interaction Inhibitor

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€214.00

€214.00


Estimated to ship on17 May 2025


Request a Bulk Order

Select a Size

Change View
10 MG
€214.00

About This Item

Empirical Formula (Hill Notation):
C13H13ClN4OS
CAS Number:
Molecular Weight:
308.79
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

€214.00


Estimated to ship on17 May 2025


Request a Bulk Order

Product Name

Fn14 Antagonist, L524-0366,

Assay

≥98% (HPLC)

Quality Level

form

powder

potency

7.12 μM Ki

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 25 mg/mL

storage temp.

2-8°C

SMILES string

C1CSCCN1C(=O)C2=CN(N=N2)C3=CC(=CC=C3)Cl

InChI

1S/C13H13ClN4OS/c14-10-2-1-3-11(8-10)18-9-12(15-16-18)13(19)17-4-6-20-7-5-17/h1-3,8-9H,4-7H2

InChI key

ZOBDTKWDYMRKPF-UHFFFAOYSA-N

General description

A triazolyl-thiomorpholinyl-methanone compound that acts as a fibroblast growth factor-inducible 14 (Fn14) antagonist via direct affinity interaction (KD = 7.12 µM) and competes against TNF-like Weak Inducer of Apoptosis (TWEAK) C-terminal/extracellular TNF homology domain (THD) for Fn14 binding (16.8% inhibition against 500 pg/mL sTWEAK; [Drug] = 25 µM). Shown to completely inhibit 100 ng/mL sTWEAK-stimulated migration of T98G glioma cells at 10 µM concentration and is more potent against sTWEAK- than TNFα-induced NF-κB activation (IC50 = 7.8 vs. 31.03 µM, respectively, in reporter assays using Fn14-NF-kB-Luc and NF-κB-Luc cells) without any apparent cytotoxicity up to 50 µM and 72 h in T98G cultures.

Biochem/physiol Actions

Primary Target
fibroblast growth factor-inducible 14 (Fn14)
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Use only fresh DMSO for reconstitution.

Other Notes

Dhruv, H., et al. 2013. J. Biol. Chem.288, 32261.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Harshil Dhruv et al.
The Journal of biological chemistry, 288(45), 32261-32276 (2013-09-24)
Deregulation of the TNF-like weak inducer of apoptosis (TWEAK)-fibroblast growth factor-inducible 14 (Fn14) signaling pathway is observed in many diseases, including inflammation, autoimmune diseases, and cancer. Activation of Fn14 signaling by TWEAK binding triggers cell invasion and survival and therefore

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service