Skip to Content
Merck
All Photos(1)

Key Documents

400110

Sigma-Aldrich

Human IgA₂, Myeloma

Sign Into View Organizational & Contract Pricing

Select a Size

500 μG
€252.00

€252.00


Estimated to ship on28 May 2025



Select a Size

Change View
500 μG
€252.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.77

€252.00


Estimated to ship on28 May 2025


Assay

≥95% (SDS-PAGE)

Quality Level

form

liquid

contains

≤0.1% sodium azide as preservative

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

shipped in

ambient

storage temp.

−20°C

target post-translational modification

unmodified

General description

Native IgA2 from human myeloma plasma. Human immunoglobulin A2, type λ serum, containing IgA2 as 98% of total IgA. Purified from plasma.

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Physical form

In 150 mM NaCl, 10 mM phosphate buffer, pH 7.4.

Preparation Note

Prepared from plasma that was shown to be nonreactive for HBsAg and antibodies to HCV and HBc and negative for HIV antibodies by certified tests.

Other Notes

Single arc by immunoelectrophoresis against antiserum to whole human sera. Variables associated with assay conditions will dictate the proper working dilution.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service