Skip to Content
Merck
All Photos(1)

Key Documents

5.08228

Sigma-Aldrich

L- Buthionine Sulfoximine

Synonym(s):

L- Buthionine Sulfoximine

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
€158.00

€158.00


Estimated to ship on30 May 2025


Request a Bulk Order

Select a Size

Change View
100 MG
€158.00

About This Item

Empirical Formula (Hill Notation):
C8H18N2O3S
CAS Number:
Molecular Weight:
222.31
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€158.00


Estimated to ship on30 May 2025


Request a Bulk Order

Assay

≥97% (TLC)

Quality Level

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

water: 50 mg/mL

storage temp.

2-8°C

InChI

1S/C8H18N2O3S/c1-2-3-5-14(10,13)6-4-7(9)8(11)12/h7,10H,2-6,9H2,1H3,(H,11,12)/t7-,14?/m0/s1

InChI key

KJQFBVYMGADDTQ-CVSPRKDYSA-N

General description

A cell-permeable, sulfoximine based compound that acts a potent, fast acting, irreversible inhibitor of g-glutamylcysteine synthetase and depletes cellular glutathione levels. Closely resembles the structure of g-glutamylphosphate cysteine adduct. The inhibition follows the first order kinetics with t1/2 = ~ 11 sec at saturating inhibitor concentration. Does not affect the activity of glutamine synthetase. Shown to improve the outcome of chemotherapy by lowering the levels of glutathione in cancer cells (IC50 = 1.9, 8.6, and 29 µM in melanoma, breast, and ovarian tumor cells, respectively).

Biochem/physiol Actions

Primary Target
gamma-glutamylcysteine synthetase

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Fruehauf, J.P., et al. 1997. Pigment Cell Res.10, 236.
OʹDwyer, P.J., et al. 1996. J. Clin. Oncol.14, 249.
Griffith, O. W., 1982. J. Biol. Chem.257, 13704.

Griffith, O. W., et al. 1979. J. Biol. Chem.254, 7558.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service