Skip to Content
Merck
All Photos(1)

Documents

506274

Sigma-Aldrich

PACOCF3

A novel Ca2+-independent and reversible inhibitor of phospholipase A₂ (PLA₂; IC₅₀ = 3.8 µM).

Synonym(s):

PACOCF3, Palmitoyl Trifluoromethyl Ketone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H31F3O
CAS Number:
Molecular Weight:
308.42
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.77

Quality Level

Assay

≥97% (TLC)

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

shipped in

wet ice

storage temp.

−20°C

InChI

1S/C17H31F3O/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16(21)17(18,19)20/h2-15H2,1H3

InChI key

MAHYXYTYTLCTQD-UHFFFAOYSA-N

General description

A novel Ca2+-independent and reversible inhibitor of phospholipase A2 (PLA2; IC50 = 3.8 µM). May also inhibit Ca2+-dependent PLA2 at much higher concentrations (IC50 = 45 µM).
A novel Ca2+-independent phospholipase A2 inhibitor (IC50 = 3.8 µM). May also inhibit Ca2+-dependent phospholipase A2 at higher concentrations (IC50 = 45 µM)

Biochem/physiol Actions

Cell permeable: no
Product does not compete with ATP.
Reversible: yes
Target IC50: 3.8 µM and 45 µM against phospholipase A2 (PLA2) and Ca2+-dependent PLA2, respectively.

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution in DMSO, aliquot and freeze (-20°C). Stock solutions are stable for up to 1 month at -20°C. Product has a very low melting point. The product will change from a solid to a liquid if held in your hand or kept at room temperature or warmer for any period of time. It is best to keep the product cool while in use by placing it in the refrigerator or on ice during experimentation. Once diluted in buffer, do not store in aqueous solutions for >1 day.
Supplied as a solution in ethanol. To change the solvent, evaporate the ethanol with a stream of nitrogen and immediately reconstitute with the solvent of choice. Soluble in DMSO (30 mg/ml). Only minimally soluble in aqueous buffers; do not use basic buffers; unstable at basic pH.

Other Notes

Conde-Frieboes, K., et al. 1996. J. Am. Chem. Soc. 118, 5519.
Lio, Y.C., et al. 1996. Biochim. Biophys. Acta 1302, 55.
Ackermann, E.J., et al. 1995. J. Biol. Chem.270, 445.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service