Skip to Content
Merck

Skip To

128136

7α, 25-dihydroxycholesterol

Synonym(s):

7α, 25-dihydroxycholesterol, EBI2 agonist, EBI2 Activator, GPR183 Activator, 7alpha,25-OHC

Sign In to View Organizational & Contract Pricing.

About This Item

Empirical Formula (Hill Notation):
C27H46O3
CAS Number:
Molecular Weight:
418.65
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.77
Assay:
≥98% (HPLC)
Form:
powder
Storage condition:
OK to freeze, protect from light


assay

≥98% (HPLC)

Quality Segment

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze, protect from light

color

white

solubility

DMSO: 25 mg/mL

shipped in

wet ice

storage temp.

−20°C

SMILES string

O[C@H]1[C@H]2[C@H]3[C@@]([C@H](CC3)[C@@H](CCCC(O)(C)C)C)(CC[C@@H]2[C@]4(CC[C@@H](CC4=C1)O)C)C

InChI key

BQMSKLCEWBSPPY-IKVTXIKFSA-N

General description

A highly potent and specific oxysterol EBI2 (GPR183) agonist with a kd of 450 pM in a saturation binding analysis, and activates EBI2 (EC50=140 pM compared with 2.1nM for its enantiomer 7b,25-OHC) in a GTP-γ assay. Dose-dependenty suppresses forskolin-induced cAMP accumulation in EBI2-expressing SK-N-MC/CRE-β galactosidase cell line with an IC50 of 2 nM, but not in control cells. Stimulates migration of LPS-activated spleen B cells and T cells in a dose-dependent manner. In addition, pharmacological inhibition of its biosynthesis in vivo by Clotrimazole, a CYP7B1inhibitor promotes the migration of adoptively transferred pre-activated B cells to the T/B boundary, mimicking the phenotype of pre-activated B cells from EBI2-deficient mice.

Packaging

Packaged under inert gas

Preparation Note

Following reconstitution, aliquot and freeze (-20°C. Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Liu, C., et al. 2011. Nature475, 519.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)


Still not finding the right product?

Explore all of our products under 7α, 25-dihydroxycholesterol


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters