Skip to Content
Merck
All Photos(1)

Documents

5.05816

Sigma-Aldrich

Xanomeline Oxalate

Synonym(s):

Xanomeline Oxalate, 3-(hexyloxy)-4-(1-methyl-1,2,5,6-tetrahydropyridin-3-yl)-1,2,5-thiadiazole, Muscarinic Acetylcholine Receptor (mAChR) Agonist, Xanomeline

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H23N3OS · xC2H2O4
CAS Number:
Molecular Weight:
281.42 (free base basis)
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥99% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 10 mM
ethanol: 25 mM

storage temp.

2-8°C

InChI

1S/C14H23N3OS.C2H2O4/c1-3-4-5-6-10-18-14-13(15-19-16-14)12-8-7-9-17(2)11-12;3-1(4)2(5)6/h8H,3-7,9-11H2,1-2H3;(H,3,4)(H,5,6)

InChI key

ZJOUESNWCLASJP-UHFFFAOYSA-N

General description

An agonist selective for muscarinic acetylcholine receptor (mAChR) subtype M1 (EC50 = 0.3, 5, 42, 52 and 92.5 nM at M1, M3, M5, M4 and M2 receptors respectively). An antipsychotic agent. Used in improving cognitive deficits and behavioral disturbances in Alzheimer′s disease and schizophrenia.

Warning

Toxicity: Standard Handling (A)

Other Notes

Wang, D., et al., 2011. Biochem. Biophys. Res. Commun.410, 229.
Si, W., et al., 2010. Neurosci. Lett.473, 115.
Stanhope, et al., 2001. J. Exp. Pharmacol. Ther.299, 782.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service