Skip to Content
Merck
All Photos(1)

Documents

PHR1063

Supelco

Residual Solvents Mixture - Class I

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Organic Volatile Impurities Class I Mix

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

suitable for USP 467
traceable to Ph. Eur. R0250000
traceable to USP 1601102

CofA

current certificate can be downloaded

concentration

in DMSO

application(s)

environmental
pharmaceutical (small molecule)

format

multi-component solution

storage temp.

2-30°C

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Residual Solvents Mixture - Class I are CRMs and Secondary Pharmaceutical Reference Standards, traceable and qualified against the corresponding Pharmacopeial standards. Sigma-Aldrich offers Certified Reference Materials (CRMs) for the analysis of Residual Solvents. A comprehensive Certificate of Analysis offers details of the methodology and results of the qualification.

Application

These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAC0266 in the slot below. This is an example certificate only and may not be the lot that you receive.

Analyte

Description

    Benzene 11 mg/mL
    Carbon tetrachloride 20 mg/mL
    1,2-Dichloroethane 27 mg/mL
    1,1-Dichloroethene 25 mg/mL
    1,1,1-Trichloroethane 50 mg/mL

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Carc. 1A - Muta. 1B - Ozone 1 - Skin Sens. 1 - STOT RE 1 - STOT RE 2 Inhalation

Target Organs

Liver,Kidney, Nose,Liver,Kidney

WGK

WGK 3

Flash Point(F)

197.6 °F

Flash Point(C)

92 °C


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

EU REACH Annex XVII (Restriction List)

EU REACH Annex XIV (Authorisation List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service