Skip to Content
Merck
All Photos(1)

Key Documents

5.04865

Sigma-Aldrich

UCOE® Expression Vector - Mouse 3.2 kb Puro Set - Novagen®

Sign Into View Organizational & Contract Pricing

Select a Size

1 SET
€815.00

€815.00


Estimated to ship on30 May 2025



Select a Size

Change View
1 SET
€815.00

About This Item

UNSPSC Code:
41105500
NACRES:
NA.51

€815.00


Estimated to ship on30 May 2025


manufacturer/tradename

Novagen®

Quality Level

storage condition

OK to freeze

storage temp.

−20°C

General description

UCOE® (Ubiquitous Chromatin Opening Element) expression technology demonstrates major improvements in gene expression in stably-transfected mammalian cells. Over 50% of UCOE derived clones have higher expression than the best non-UCOE derived vectors. UCOE vectors may be used to express antibodies, receptors, enzymes, cytokines, and other similar sized biological material. UCOE technology is convenient and easy to use, because it integrates with industry standard platforms (e.g. CHO cells, CMV promoters, plasmids, transfection agents, media, etc...).

The set includes following components:
• 1 x 10 µg DNA
• 1 x 10 µg DNA (Negative control)
Antibiotic resistance: Puromycin




THIS PRODUCT IS ONLY AVAILABLE FOR SALE TO ACADEMIC INSTITUTIONS OR NOT-FOR-PROFIT ENTITIES FOR RESEARCH USE. FOR INFORMATION ON COMMERCIAL LICENSING OF THE UCOE TECHNOLOGY, INCLUDING TO COMMERCIAL ENTITIES, PLEASE CONTACT Marie Azzaria ([email protected]) or Jamie Hoberg ([email protected]).

Application

UCOE (Ubiquitous Chromatin Opening Elements) technology is a breakthrough solution for sustained recombinant protein production and cell line development in mammalian protein expression systems.

Warning

Toxicity: Standard Handling (A)

Legal Information

NOVAGEN is a registered trademark of Merck KGaA, Darmstadt, Germany
UCOE is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service