Skip to Content
Merck

1.00193

SAFC

Caprylic acid (Octanoic acid)

EMPROVE® EXPERT, Ph. Eur., NF

Pharma Manufacturing

Synonym(s):

Caprylic acid (Octanoic acid)

Sign Into View Organizational & Contract Pricing

Select a Size

2.5 L
€323.00
25 L
€2,974.00
200 L
Inquire

€323.00


Estimated to ship on22 May 2025


Request a Bulk OrderRequest a Sample

Select a Size

Change View
2.5 L
€323.00
25 L
€2,974.00
200 L
Inquire

About This Item

Linear Formula:
CH3(CH2)6COOH
CAS Number:
Molecular Weight:
144.21
MDL number:
UNSPSC Code:
12352106
EC Index Number:
204-677-5

€323.00


Estimated to ship on22 May 2025


Request a Bulk OrderRequest a Sample

Agency

NF
Ph. Eur.

Quality Level

vapor pressure

0.05 hPa ( 20 °C)

product line

EMPROVE® EXPERT

form

liquid

autoignition temp.

>300 °C

expl. lim.

1 % (v/v)

pH

4 (20 °C, 0.2 g/L in H2O)

bp

237 °C/1013 hPa

mp

16 °C

transition temp

flash point 130 °C

solubility

0.68 g/L

density

0.91 g/cm3 at 20 °C

application(s)

liquid formulation
pharma/biopharma processes
pharmaceutical

storage temp.

15-25°C

SMILES string

[O-]C(=O)CCCCCCC.[H+]

InChI

1S/C8H16O2/c1-2-3-4-5-6-7-8(9)10/h2-7H2,1H3,(H,9,10)

InChI key

WWZKQHOCKIZLMA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Finding the right excipient that matches your needs as well as regulatory demands can be a complicated challenge in formulation. With our application know-how and regulatory expertise, we support you in every step of development, scale-up, and production.
As part of our Emprove® Program, our raw materials are offered with extensive documentation facilitating compliance of your pharma and biopharma product, full supply chain transparency and risk mitigation. Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.

Application

Caprylic Acid also known as octanoic acid is widely used in the biopharma industry as a precipitation agent especially in purification of plasma derived therapeutics.
Due to its low microbial and endotoxin limits, Caprylic acid (Octanoic acid) Emprove® Expert is especially suitable for high-risk applications.

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1C

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

>230.0 °F - closed cup

Flash Point(C)

> 110 °C - closed cup


What is the Emprove® Program?

The Emprove® Program is a system providing comprehensive and thorough documentation of our filters and single-use components, pharma raw materials, and starting materials. Four document types are included with an Emprove® Program subscription:

Material Qualification Dossier

Free of charge - Sign in to download

Quality Management Dossier (Open Access)

Free of charge - Sign in to download

Operational Excellence Dossier

Available by subscription or for a fee

Operational Excellence Dossier

Available by subscription or for a fee

Learn more about the benefits an Emprove® Program subscription can provide
Why are these documents unavailable?
Emprove® Dossiers contain information intended to support the registration process and risk assessment activities of pharmaceutical producers and contain confidential information. Please contact us if you would like access.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service